Skip to Content
Merck
All Photos(1)

Key Documents

572660

Sigma-Aldrich

SU11652

A cell-permeable pyrrole-indolinone compound that acts as a potent, reversible, and ATP-competitive tyrosine kinase receptor and angiogenic inhibitor.

Synonym(s):

SU11652, 5-[(Z)-(5-Chloro-2-oxo-1,2-dihydro-3H-indol-3-ylidene)methyl]-N-[2-(diethylamino)ethyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide, PDGFR Tyrosine Kinase Inhibitor VIII, VEGFR Tyrosine Kinase Inhibitor XVII, VEGFR2 Kinase Inhibitor XIV

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$631.00

$631.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 μG
$631.00

About This Item

Empirical Formula (Hill Notation):
C22H27ClN4O2
CAS Number:
Molecular Weight:
414.93
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$631.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 4 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1cc2c(cc1)NC(=O)\C\2=C/c3[nH]c(c(c3C)C(=O)NCCN(CC)CC)C

InChI

1S/C22H27ClN4O2/c1-5-27(6-2)10-9-24-22(29)20-13(3)19(25-14(20)4)12-17-16-11-15(23)7-8-18(16)26-21(17)28/h7-8,11-12,25H,5-6,9-10H2,1-4H3,(H,24,29)(H,26,28)/b17-12-

InChI key

XPLJEFSRINKZLC-ATVHPVEESA-N

General description

A cell-permeable pyrrole-indolinone compound that acts as a potent, reversible, and ATP-competitive tyrosine kinase receptor and angiogenic inhibitor that exhibits greater selectivity for PDGFRβ (IC50 = 3 nM), VEGFR2 (IC50 = 27 nM), FGFR1 (IC50 = 170 nM), and Kit family members (IC50 = ~ 10-500 nM) over EGFR (IC50 >20 µM). Reported to display anti-proliferative and pro-apoptotic properties in tumor cells.
A cell-permeable, potent, reversible, and ATP-competitive inhibitor of tyrosine kinase activity of PDGFRβ (IC50 = 3 nM), VEGFR2 (IC50 = 27 nM), FGFR1 (IC50 = 170 nM), and Kit family members (IC50 ~10-500 nM). Does not effect the activity of EGFR (IC50 >20 µM). Reported to display anti-proliferative, antiangiogenic and pro-apoptotic properties in tumor cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDGFRB
Product competes with ATP.
Reversible: yes
Target IC50: 3 nM, 27 nM, 170 nM against PDGFRβ, VEGFR2, and FGFR1, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Heryanto, B., et al. 2003. Reproduction125, 334.
Sun, L., et al. 2003. J. Med. Chem.46, 1116.
Liao, A.T., et al. 2002. Blood100, 585.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Biggi et al.
Journal of neurochemistry, 152(1), 136-150 (2019-07-03)
The vast majority of therapeutic approaches tested so far for prion diseases, transmissible neurodegenerative disorders of human and animals, tackled PrPSc , the aggregated and infectious isoform of the cellular prion protein (PrPC ), with largely unsuccessful results. Conversely, targeting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service