Skip to Content
Merck
All Photos(1)

Key Documents

25649

Sigma-Aldrich

2,2,5,7,8-Pentamethyl-chromane-6-sulfonyl chloride

≥97.0% (HPLC)

Synonym(s):

3,4-Dihydro-2,2,5,7,8-pentamethyl-2H-1-benzopyran-6-sulfonyl chloride, Pmc-Cl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$259.00

$259.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$259.00

About This Item

Empirical Formula (Hill Notation):
C14H19ClO3S
CAS Number:
Molecular Weight:
302.82
Beilstein:
4488701
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$259.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (HPLC)

form

powder

mp

77-82 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Cc1c(C)c(c(C)c2CCC(C)(C)Oc12)S(Cl)(=O)=O

InChI

1S/C14H19ClO3S/c1-8-9(2)13(19(15,16)17)10(3)11-6-7-14(4,5)18-12(8)11/h6-7H2,1-5H3

InChI key

UXUOVYKDMGFUDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent to introduce the acid labile arginine side-chain protection Pmc[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service