Skip to Content
Merck
All Photos(1)

Key Documents

557364

Sigma-Aldrich

Roscovitine

InSolution, ≥95%, inhibitor of cyclin-dependent kinases (CDKs)

Synonym(s):

InSolution Roscovitine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$360.00

$360.00


Estimated to ship on30 May 2025



Select a Size

Change View
5 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C19H26N6O
Molecular Weight:
354.45
UNSPSC Code:
12352200
NACRES:
NA.77

$360.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A potent and selective inhibitor of cyclin-dependent kinases (Cdks). Exhibits greater inhibitory potency compared to olomoucine (Cat. No. 495620). Inhibits p34 Cdc2/cyclin B (IC50 = 650 nM), p33 Cdk2/cyclin A (IC50 = 700 nM), p33 Cdk2/cyclin E (IC50 = 700 nM), and p33 Cdk2/p35 (IC50 = 200 nM) by competing for the ATP binding domain of these kinases. Exhibits very low sensitivity towards related kinases, such as Erk1 and Erk2 (IC50 = 34 µM and 14 µM, respectively). Roscovitine does not significantly affect the activity of other protein kinases even at 100 µM and compared to olomoucine, roscovitine displays increased anti-mitotic activity at the G1/S and G2/M phases of the cell cycle.

Biochem/physiol Actions

Cell permeable: no
Primary Target
p34cdk1/cyclin B
Product competes with ATP.
Reversible: yes
Target IC50: 650 nM, 700 nM, 700 nM, and 200 nM against p34cdk1/cyclin B, p33cdk2/cyclin A, p33cdk2/cyclin E, and p33cdk5/p35, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/282 µl) solution of Roscovitine (Cat. No. 557360) in DMSO.

Reconstitution

Following initial use, aliquot and freeze (-20°C). Once opened, the DMSO aliquots are stable for up to 6 months at -20°C.

Other Notes

Meijer, L. 1996. Trends Cell Biol.6, 393.
Rudolph, B., et al. 1996. EMBO J.15, 3053.

Legal Information

Sold under license of U.S. Patent 6,316,456.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Juan Cheng et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(46) (2021-11-10)
What determines the rate at which a multicellular organism matures is a fundamental question in biology. In plants, the decline of miR156 with age serves as an intrinsic, evolutionarily conserved timer for the juvenile-to-adult phase transition. However, the way in
Homology directed telomere clustering, ultrabright telomere formation and nuclear envelope rupture in cells lacking TRF2B and RAP1.
Rai, et al.
Nature Communications, 14, 2144-2144 (2023)
Manzar Hossain et al.
Molecular cell, 81(9), 1951-1969 (2021-03-25)
The initiation of DNA replication involves cell cycle-dependent assembly and disassembly of protein complexes, including the origin recognition complex (ORC) and CDC6 AAA+ ATPases. We report that multiple short linear protein motifs (SLiMs) within intrinsically disordered regions (IDRs) in ORC1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service