Skip to Content
Merck
All Photos(1)

Key Documents

539211

Sigma-Aldrich

Protein Methyltransferase Inhibitor, AMI-5

The Protein Methyltransferase Inhibitor, AMI-5, also referenced under CAS 17372-87-1, controls the biological activity of Protein Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protein Methyltransferase Inhibitor, AMI-5, Disodium-2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate, trihydrate, A(K)MI-5, PRMT Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H6Br4Na2O5 · 3H2O
CAS Number:
Molecular Weight:
854.00
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.54
Product 539211 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥90%

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

red-brown

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].Brc1c2[o][c]3[c](c(c2cc([c]1=O)Br)c4c(cccc4)C(=O)[O-])=CC(=C(C=3Br)[O-])Br

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

General description

A xanthenyl compound that inhibits the activities of protein arginine methyltransferases (PRMT) Hmt1p, PRMT1/3/4/6 (IC50 against yeast Hmt1p & human PRMT1 = 0.78 & 1.41 µM, respectively), as well as histone lysine methyltransferases (HMTases) SET7 and DOT1 in an AdoMet/SAM- (S-adenosylmethionine) competitive manner. Although SAM-competitive inhibitors generally exhibit broad activity against and are non-selective toward most SAM-utilizing enzymes, AMI-5 is known to be inactive against at least two HMTases (Suv39H1 & Suv39H2).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Cheng, D., et al. 2004. J. Biol. Chem.279, 23892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

376.9 °F - Pensky-Martens closed cup

Flash Point(C)

191.6 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Wohlfromm et al.
Oncogene, 43(25), 1955-1971 (2024-05-11)
Procaspase-8 is a key mediator of death receptor (DR)-mediated pathways. Recently, the role of post-translational modifications (PTMs) of procaspase-8 in controlling cell death has received increasing attention. Here, using mass spectrometry screening, pharmacological inhibition and biochemical assays, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service