Skip to Content
Merck
All Photos(1)

Key Documents

525185

Sigma-Aldrich

Phospholipase C Activator, m-3M3FBS

The Phospholipase C Activator, m-3M3FBS, also referenced under CAS 200933-14-8, controls the biological activity of Phospholipase C. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Phospholipase C Activator, m-3M3FBS, m-3M3FBS, 2,4,6-Trimethyl-N-( m-3-trifluoromethylphenyl)benzenesulfonamide, m-3M3FBS, 2,4,6-Trimethyl-N-(m-3-trifluoromethylphenyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16F3NO2S
CAS Number:
Molecular Weight:
343.36
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Product 525185 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 7 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)N[S](=O)(=O)c2c(cc(cc2C)C)C

InChI

1S/C16H16F3NO2S/c1-10-7-11(2)15(12(3)8-10)23(21,22)20-14-6-4-5-13(9-14)16(17,18)19/h4-9,20H,1-3H3

InChI key

ZIIUUSVHCHPIQD-UHFFFAOYSA-N

General description

A cell-permeable and specific activator of phospholipase C (PLC). Reported to stimulate PLC-mediated intracellular Ca2+ release, inositol phosphate production, and superoxide generation in various cell types. Activates all PLC isotypes (β2, β3, γ1, &gamma2, and δ1) in vitro, but does not exhibit any effect on heterotrimeric G proteins, PI 3-kinase, or phospholipase D.
A novel sulfonamide compound that acts as a cell-permeable and specific activator of phospholipase C (PLC). Shown to stimulate the PLC-mediated intracellular Ca2+ release, inositol phosphates production, and superoxide generation in various cell types. It activates all PLC isotypes (β2, β3, γ1, γ2, and δ1) in vitro, but not heterotrimeric G proteins, PI 3-kinase, or PLD.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Phospholipase C (PLC)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Bae, Y.S., et al. 2003. Mol. Pharmacol.63, 1043.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouhannad Malek et al.
The Journal of biological chemistry, 298(3), 101740-101740 (2022-02-20)
Lysosomes serve as dynamic regulators of cell and organismal physiology by integrating the degradation of macromolecules with receptor and nutrient signaling. Previous studies have established that activation of the transcription factor EB (TFEB) and transcription factor E3 (TFE3) induces the
Mouhannad Malek et al.
Nature communications, 12(1), 2673-2673 (2021-05-13)
Vesicular traffic and membrane contact sites between organelles enable the exchange of proteins, lipids, and metabolites. Recruitment of tethers to contact sites between the endoplasmic reticulum (ER) and the plasma membrane is often triggered by calcium. Here we reveal a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service