Skip to Content
Merck
All Photos(1)

Key Documents

524653

Millipore

PBS-TWEEN® Tablets

tablet, 140 mM NaCl, 3 mM KCl, 0.05% TWEEN 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C

Synonym(s):

PBS, Phosphate Buffered Saline-TWEEN® 20 Tablets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$176.00

$176.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$176.00

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

$176.00


Available to ship on31 March 2025Details


Request a Bulk Order

Product Name

PBS-TWEEN® Tablets, Dissolve one tablet in 1 liter of deionized water to yield 140 mM NaCl, 3 mM KCl, 0.05% TWEEN 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C. Note: 1 each = 10 tablets.

Quality Level

form

tablet

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

deionized water: 1 tablet/liter

shipped in

ambient

storage temp.

15-25°C

General description

Developed for use in immunoassay applications. Dissolving one tablet in 1 liter of deionized H2O yields 140 mM NaCl, 2.7 mM KCl, 0.05% TWEEN® 20 detergent, and 10 mM phosphate buffer, pH 7.4 at 25°C. Note: 1 each = 10 tablets.
Dissolve one tablet in 1 liter of deionized water to yield 140 mM NaCl, 3 mM KCl, 0.05% TWEEN® 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C. Note: 1 each = 10 tablets.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C) or filter sterilized and store at room temperature (20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. Is this product sterile/usable in the context of microbiological analysis ?

    1 answer
    1. The product is not sterile. No protocol is offered concerning its use in microbiological analyses.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service