Skip to Content
Merck
All Photos(2)

Key Documents

524619

Sigma-Aldrich

PIP3 Antagonist II, DM-PIT-1

The PIP3 Antagonist II, DM-PIT-1, also referenced under CAS 701947-53-7, controls the biological activity of PIP3.

Synonym(s):

PIP3 Antagonist II, DM-PIT-1, N-​(((2-Hydroxy-​5-​nitrophenyl)amino)thioxomethyl)-​3,​5-​dimethyl-​benzamide, N-((2-Hydroxy-5-nitrophenyl)carbamothioyl)-3,5-dimethylbenzamide, Akt Inhibitor XXII, PDK1 Inhibitor VI

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$280.00

$280.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$280.00

About This Item

Empirical Formula (Hill Notation):
C16H15N3O4S
CAS Number:
Molecular Weight:
345.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$280.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(C=CC([N+]([O-])=O)=C1)=C1NC(NC(C2=CC(C)=CC(C)=C2)=O)=S

InChI

1S/C16H15N3O4S/c1-9-5-10(2)7-11(6-9)15(21)18-16(24)17-13-8-12(19(22)23)3-4-14(13)20/h3-8,20H,1-2H3,(H2,17,18,21,24)

InChI key

CGWBCAQMXJFWBU-UHFFFAOYSA-N

General description

A cell-permeable benzoylthiourea compound that is shown to compete against PIP3 for binding PH (pleckstrin-homology) domains of Akt1 (IC50 ≥31 µM using recombinant human Akt1 aa 1-123), ARNO, GRP1, and PKD1, while lacking activity against PIP3 Btk PH domain ineraction, PI-4,5-P2 (Cat. Nos. 524644) PLC-δ PH domain binding, or PI-3,4-P2 TAPP1/2 PH domains binding, resulting in effective blockage of PIP3-dependent PI3K-PDK1-Akt signaling pathway activation in human glioblastoma U87MG cultures (25 to 100 µM for 3 d) and (5 min 100 ng/ml) PDGF-induced Akt and GRP membrane translocation (by 85% and 70%, respectively, with 1 h 100 µM pretreatment) in serum-starved human breast cancer SUM159 cells, while being inactive against PDGF-induced Btk translocation or (5 min 250 ng/ml) PMA-induced PLC-δ and TAPP1/2 translocations. Although DM-PIT-1 can be administered as a DMSO stock for effective culture treatments, incorporating DM-PIT-1 into PEG-PE mixed micelles is reported to enhance its solubility (up to 1 mM), and presumably delivery efficiency (57% vs. 67% U87MG viability after 24 h 50 µM drug treatment, respectively, with or without PEG-PE encapsulation), making large dosage delivery possible for more effective in vivo administrations (5% vs. 41% of control 4T1 tumor size in mice after 8 daily i.v. dosages of 1 mg/kg micelles-formulated or 0.4 mg/kg free drug, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Miao, B., et al. 2010. Proc. Natl. Acad. Sci. USA107, 20126.
Skidan, I., et al. 2009. Drug Deliv.16, 45.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Skidan et al.
Drug delivery, 16(1), 45-51 (2009-06-27)
The purpose of this study was to develope and characterize a micellar formulations of N-{[(2-hydroxy-5- nitrophenyl)amino]carbonothioyl}-3,5-dimethylbenzamide (DM-PIT-1)-a new small molecule non-lipid antagonist of phopshotidylinositol-3.4.5-triphopshate and inhibitor of the PI3-kinase pathway. Micelle-forming PEG(2000)-PE was used to solubilize DM-PIT-1. To improve the
Benchun Miao et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(46), 20126-20131 (2010-11-03)
The PI3-kinase (PI3K) pathway regulates many cellular processes, especially cell metabolism, cell survival, and apoptosis. Phosphatidylinositol-3,4,5-trisphosphate (PIP3), the product of PI3K activity and a key signaling molecule, acts by recruiting pleckstrin-homology (PH) domain-containing proteins to cell membranes. Here, we describe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service