Skip to Content
Merck
All Photos(1)

Key Documents

521275

Sigma-Aldrich

PDK1/Akt/Flt Dual Pathway Inhibitor

The PDK1/Akt/Flt Dual Pathway Inhibitor, also referenced under CAS 331253-86-2, controls the biological activity of PDK1/Akt/Flt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PDK1/Akt/Flt Dual Pathway Inhibitor, 6H-Indeno[1,2-e]tetrazolo[1,5-b][1,2,4]triazin-6-one & 10H-Indeno[2,1-e]tetrazolo[1,5-b][1,2,4]triazin-10-one, KP372-1, Akt Inhibitor XXI, PDK1 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$560.00

$560.00


Estimated to ship on15 May 2025



Select a Size

Change View
5 MG
$560.00

About This Item

Empirical Formula (Hill Notation):
C10H4N6O
CAS Number:
Molecular Weight:
224.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$560.00


Estimated to ship on15 May 2025


Quality Level

Assay

≥98% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21nnnc2nc3c(n1)c4c(cccc4)C3=O

InChI

1S/C10H4N6O/c17-9-6-4-2-1-3-5(6)7-8(9)11-10-12-14-15-16(10)13-7/h1-4H

InChI key

SSNMLKNJEUSPDY-UHFFFAOYSA-N

General description

A cell-permeable compound that selectively induces apoptosis in AML (Acute Myelogenous Leukemia) with little effect on normal CD34+ AML progenitor cells. Shown to directly inhibit both PDK1 and Akt activities in in vitro kinase assays in a dose-dependent manner and block cellular phosphorylation of Akt at both Ser473 and Thr308. The dual inhibition nature against both PDK1/Akt and Flt3/PIM signaling pathways allows effective killing of AML cells (Average IC50 = 1.05, 1.91, and 0.43 µM for AML with wild-type Flt3, single mutant ITD/D835, and double mutant Flt3-ITD-TDK, respectively) that are otherwise resistant to inhibitors that target only the PDK1/Akt pathway. CAS. No. 331253-86-2 and 329710-24-9.
CAS. Nos. 331253-86-2 and 329710-24-9 (two isomers)

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Both PDK1 and Akt activities
Product does not compete with ATP.
Reversible: no
Target IC50: 1.05, 1.91, and 0.43 µM for AML with wild-type Flt3, single mutant ITD/D835, and double mutant Flt3-ITD-TDK, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Zeng, Z., et al. 2006. Cancer Res.66, 3737.
Koul, D., et al. 2006. Mol. Cancer Ther.5, 637.
Mandal. M., et al. 2006. Oral Oncol.42, 430.
Mandal. M., et al. 2005. Br. J. Cancer92, 1899.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service