Skip to Content
Merck
All Photos(2)

Key Documents

521238

Sigma-Aldrich

PDGFRβ/RAF Kinase Inhibitor

The PDGFRβ/RAF Kinase Inhibitor controls the biological activity of PDGFRβ/RAF Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PDGFRβ/RAF Kinase Inhibitor, PDGFR Tyrosine Kinase Inhibitor XXII, 6-(4-(5-(3-(Trifluoromethyl)phenylamino)-4 H-1,2,4-triazol-3-yl)phenoxy)-2-(methylthio)pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16F3N7OS
CAS Number:
Molecular Weight:
459.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

NC1=CC(OC2=CC=C(C3=NN=C(NC4=CC(C(F)(F)F)=CC=C4)N3)C=C2)=NC(SC)=N1

General description

An orally bioavailable amino-triazole compound that is shown to bind to the inactive conformations of both PDGFRβ and Raf kinases (allosteric, Type II inh), and acts as a potent inhibitor of Flt3, Kit, PDGFRα and PDGFRβ (Kd = 52, 170, 300 and 520 nM, respectively) with selectivity over B-Raf, p38α, FGFR2 and VEGFR2 (Kd >10 µM) in a multi-kinase panel. Completely blocks the phosphorylations of MEK-1/2 S217/S221, ERK-1/2 T202/Y204 in FGF-stimulated HUVECs, PDGFRβ in PDGF-BB-stimulated VSMCs, ERK-1/2 T202/Y204 in 1205Lu-B Raf V600E cells, and C-Raf S338 at 10 µM. Reduces FGF-induced angiogenesis in mouse Matrigel model (50 mg/kg, i.p., bid) and selectively induces apoptosis in the endothelium; further, suppresses orthotopic renal cell carcinoma growth in mice (100 mg/kg, p.o., qd).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Murphy, E.A., et al. 2010. Proc. Natl. Acad. Sci. USA107, 4299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service