Skip to Content
Merck
All Photos(1)

Key Documents

506163

Sigma-Aldrich

p38 MAP Kinase Inhibitor VIII

The p38 MAP Kinase Inhibitor VIII, also referenced under CAS 321351-00-2, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p38 MAP Kinase Inhibitor VIII, (4-((2-Amino-4-bromophenyl)amino)-2-chlorophenyl)-(2-methylphenyl)methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$238.00

$238.00


Estimated to ship on19 May 2025



Select a Size

Change View
5 MG
$238.00

About This Item

Empirical Formula (Hill Notation):
C20H16BrClN2O
CAS Number:
Molecular Weight:
415.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$238.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

ethanol: 15 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1cc(c(cc1)Nc2cc(c(cc2)C(=O)c3c(cccc3)C)Cl)N

InChI

1S/C20H16BrClN2O/c1-12-4-2-3-5-15(12)20(25)16-8-7-14(11-17(16)22)24-19-9-6-13(21)10-18(19)23/h2-11,24H,23H2,1H3

InChI key

HDCLCHNAEZNGNV-UHFFFAOYSA-N

General description

A cell-permeable aminobenzophenone compound that acts as a potent, ATP-binding pocket-targeting inhibitor against p38α and p38β2 (82% and 93% inhibition with 1 µM inhibitor, respectively; IC50 of p38α inhibition = 40 nM) with little activity against a panel of 57 other kinases, including p38γ, p38δ, Erk1/2, and JNK1. Shown to inhibit LPS-induced cytokine productions in human PBMC′s in vitro and exhibit in vivo efficacy in both acute and chronic murine models of skin inflammation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ottosen, E.R., et al. 2003. J. Med. Chem.46, 5651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service