Skip to Content
Merck
All Photos(1)

Key Documents

559397

Sigma-Aldrich

SB 202190

InSolution, ≥98%, p38 MAP kinase inhibitor

Synonym(s):

InSolution SB 202190, InSolution p38 MAP Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$322.00

$322.00


Estimated to ship on24 May 2025



Select a Size

Change View
1 ML
$322.00

About This Item

Empirical Formula (Hill Notation):
C20H14FN3O
CAS Number:
Molecular Weight:
331.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$322.00


Estimated to ship on24 May 2025


Quality Level

Assay

≥98% (HPLC)

form

liquid

potency

16 nM IC50
16 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)C2=C(NC(=C4C=CC(=O)C=C4)N2)c3ccncc3

InChI

1S/C20H14FN3O/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(25)8-4-15/h1-12,23-24H

InChI key

NJNKPVPFGLGHPA-UHFFFAOYSA-N

General description

A potent and cell-permeable inhibitor of p38 MAP kinase. Inhibits p38 phosphorylation of myelin basic protein (MBP) with no effect on the activity of the ERK or JNK MAP kinase subgroups. Also inhibits the kinase activity of p38β (IC50 = 350 nM) and p38 phosphorylation of activating transcription factor 2 (ATF-2; IC50 = 280 nM). Blocks LPS-induced TNF-α and interleukin biosynthesis. Reported to block LDL expression in Hep52 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
P38β
Product competes with ATP.
Reversible: yes
Target Ki: 350 nM against the kinase activity of p38β; 280 nM against p38 phosphorylation of activating transcription factor 2 (ATF-2)

Packaging

Packaged under inert gas
Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Irritant (B)

Physical form

A 1 mg/ml solution of SB 202190 (Cat. No. 559388) in anhydrous DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Ajizian, S.J., et al. 1999. J. Infect. Dis.179, 939.
Singh, R.P., et al. 1999. J. Biol. Chem.274, 19593.
Wang, S.W., et al. 1999. Inflamm. Res.48, 533.
Gallagher, T.F., et al. 1997. Bioorg. Med. Chem.5, 49.
Jiang, Y., et al. 1996. J. Biol. Chem.271, 17920.
Kramer, R.M., et al. 1996. J. Biol. Chem.271, 27723.
Li, Z., et al. 1996. Biochem. Biophys. Res. Commun.228, 334.
Lee, L.C., et al. 1994. Nature372, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Cui et al.
Cell biology international, 44(9), 1811-1819 (2020-05-07)
Multiple sclerosis (MS) is an autoimmune disease of the central nervous system that is caused by the interaction of genetic and environmental factors. Current studies have shown that Fc-receptor like-3 (FCRL3) is closely related to MS, but the specific role
Nisha R Pawar et al.
The Journal of cell biology, 222(11) (2023-09-22)
The transmembrane serine protease matriptase is a key regulator of both barrier-disruptive and protective epithelial cell-cell interactions. Elevated matriptase is a consistent feature of epithelial ovarian cancers (OvCa), where multicellular spheroids shed from the primary tumor into the peritoneal cavity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service