Skip to Content
Merck
All Photos(1)

Key Documents

5.06072

Sigma-Aldrich

IWP-2

≥99% (HPLC), liquid, Wnt antagonist, Calbiochem

Synonym(s):

InSolution Wnt Antagonist II, IWP-2, N-(6-Methylbenzo[d]thiazol-2-yl)-2-(4-oxo-3-phenyl-3,4,6,7-tetrahydrothieno[3,2-d]pyrimidin-2-ylthio)acetamide, Wnt Pathway Inhibitor II InSolution, Inhibitor of Wnt Production-2, Porcn Inhibitor I InSolution, InSolution Wnt Antag

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$191.00

$191.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C22H18N4O2S3
CAS Number:
Molecular Weight:
466.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$191.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Product Name

IWP-2, InSolution, ≥99%, 10 mM in DMSO, A cell-permeable benzothiazolyl-acetamide compound, Wnt Antagonist II

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

[s]1c2c(nc1NC(=O)CSc3[n]([c](c5c(n3)CCS5)=O)c4ccccc4)ccc(c2)C

InChI

1S/C22H18N4O2S3/c1-13-7-8-15-17(11-13)31-21(23-15)25-18(27)12-30-22-24-16-9-10-29-19(16)20(28)26(22)14-5-3-2-4-6-14/h2-8,11H,9-10,12H2,1H3,(H,23,25,27)

InChI key

WRKPZSMRWPJJDH-UHFFFAOYSA-N

General description

A cell-permeable benzothiazolyl-acetamide compound that inhibits the cellular Wnt processing and secretion via selective blockage of MBOAT (membrane-bound O-acyltranferase) family member Porcn- (Porcupine) mediated Wnt palmitoylation. IWP-2 does not affect Wnt/β-catenin signaling pathway in general and, unlike IWR-1-endo (Cat. No. 681669), displays no effect against Wnt-stimulated cellular responses.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Porcn
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 10 mM (5 mg/1.07 ml) sterile-filtered solution of Wnt Antagonist II, IWP-2 (Cat. No. 681671) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C. Warming to 60°C may be required upon initial thaw.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service