Skip to Content
Merck
All Photos(1)

Key Documents

918849

Sigma-Aldrich

Zirconium oxide

monoclinic, <10 μm particle size, 99.6%

Synonym(s):

Zirconium(IV) oxide, Zirconia

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$419.00

$419.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$419.00

About This Item

Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$419.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

99.6%

form

powder

particle size

<10 μm

bp

5000 °C (lit.)

mp

2700 °C (lit.)

density

5.89 g/mL at 25 °C (lit.)

SMILES string

O=[Zr]=O

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zirconium oxide (ZrO2) is a white powder characterized by its monoclinic crystal structure and a particle size of less than 10 microns. This material appears as a white powder and is known for its high stability and resistance to corrosion. Zirconium oxide combines excellent mechanical properties with thermal stability, making it suitable for a wide range of applications, including ceramics, thermal barrier coatings, and dental materials. Its unique properties enable its use in environments requiring durability and high performance.

Application

Zirconium(IV) oxide (ZrO2) which is also known as zirconia is a ceramic nanoparticle that can be used as a nano-filler. ZrO2 process yields a consistently high purity zirconia composed of monoclinic phase particles. The low surface area and unique spheroid-shaped particles find applications in ceramics, high-temperature fillers, sensors, and catalysts.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service