Skip to Content
Merck
All Photos(1)

Key Documents

5.04377

Sigma-Aldrich

AMPK Signaling Activator XIII, ZLN005

Synonym(s):

AMPK Signaling Activator XIII, ZLN005, 2-(4- tert-Butylphenyl)-1H-benzo[d]imidazole, 2-(4-(2-Methyl-2-propanyl)phenyl)-1H-benzimidazole, Luciferase Inhibitor IV, 2-(4-tert-Butylphenyl)-1H-benzo[d]imidazole, 2-(4-(2-Methyl-2-propanyl)phenyl)-1H-benzimidazole, Luciferase Inhibitor IV

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$293.00

$293.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
25 MG
$293.00

About This Item

Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$293.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pink-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1c2c(nc1c3ccc(cc3)C(C)(C)C)cccc2

InChI

1S/C17H18N2/c1-17(2,3)13-10-8-12(9-11-13)16-18-14-6-4-5-7-15(14)19-16/h4-11H,1-3H3,(H,18,19)

InChI key

LQUNNCQSFFKSSK-UHFFFAOYSA-N

General description

A cell-permeable benzimidazole that, in addition to its inhibitory potency against luciferase activity (IC50<10 nM), is reported to activate AMPK signaling by increasing cellular ADP/ATP ratio via mitochondria respiration uncoupling in a cell type-specific manner, effectively activating AMPK & PGC-1α in muscle, but not liver, cell types both in cultures (5-20 µM & 1-24 h in rat L6 myotubes; no effect in primary rat hepatocytes) in vitro and in db/db mice (gastrocnemius/calf muscle, but not liver tissue; 15 mg/kg/day via p.o. for 6 wks) in vivo. ZLN005 is orally available in mice (plasma Cmax & Tmax = 3.7 µM & 15 min, respectively, following single 15 mg/kg oral dosage to db/db mice) and is efficacious in improving glucose (1.5 g/kg i.p.) tolerance among 4-wk-treated and insulin (1 unit/kg) tolerance among 5-wk treated db/db mice (by 14% and 18%, respectively; 15 mg/kg/day via p.o.) in vivo without any apparent effects on body weight or food intake among db/db or non-diabetic controls.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
increases cellular ADP/ATP ratio via mitochondria respiration uncoupling
Secondary Target
luciferase activity inhibitor

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, L.N., et al. 2013. Diabetes.62, 1297.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service