Skip to Content
Merck
All Photos(1)

Key Documents

NS01L

Sigma-Aldrich

Normal Rabbit Serum

This Normal Rabbit Serum is validated for use in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Rabbit Serum.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$128.00

$128.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$128.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
biological source:
rabbit
form:
lyophilized
shipped in:
ambient

$128.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

form

lyophilized

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Normal carrier sera from healthy, non-immunized rabbits.

Application

ELISA (see comments)

Flow Cytometry (see comments)

Immunoblotting (see comments)

Immunofluorescence (see comments)

Immunohistochemistry (see comments)

Immunoprecipitation (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted in 250 mM NaCl, 10 mM PBS, pH 7.6.

Reconstitution

Reconstitute the lypohilized diluted serum in 1 ml sterile distilled water with occasional gentle mixing for at least 2 h at 4°C. Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) with 0.1% azide for short-term storage. Avoid freeze/thaw cycles of solutions.

Other Notes

Normal sera are provided to be used as blocking agents in immunohistochemical procedures or as negative controls for undiluted serum/rabbit polyclonal antibodies from the Calbiochem brand. For use as a negative control, perform experiments in parallel using primary antibody and normal serum at the same concentrations. A signal obtained with the normal serum indicates a nonspecific interaction; any additional signal with the primary antibody indicates a specific antibody-antigen interaction. This antibody has also been reported to work for ELISA, flow cytometry, immunoblotting, immunofluorescence, immunohistochemistry, and immunoprecipitation.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service