Skip to Content
Merck
All Photos(1)

Key Documents

5.04173

Sigma-Aldrich

MAT2A Inhibitor II, FIDAS-5

Synonym(s):

MAT2A Inhibitor II, FIDAS-5, FIDAS5, Methionine S-Adenosyltransferase 2A Inhibitor II, (E)-4-(2-Chloro-6-fluorostyryl)-N-methylaniline, Methionine S-Adenosyltransferase 2A Inhibitor II, FIDAS5, (E)-4-(2-Chloro-6-fluorostyryl)-N-methylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$231.00

$231.00


Estimated to ship on21 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$231.00

About This Item

Empirical Formula (Hill Notation):
C15H13ClFN
CAS Number:
Molecular Weight:
261.72
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$231.00


Estimated to ship on21 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Fc1c(c(ccc1)Cl)\C=C\c2ccc(cc2)NC

InChI key

KXVXICBOMOGFMH-JXMROGBWSA-N

General description

A cell-permeable fluorinated N,N-dialkylaminostilbene (FIDAS) analogue of FIDAS-3 that is >2-fold more potent than FIDAS-3 in inhibiting MAT2A-catalyzed SAM (S-adenosylmethionine) synthesis both in cell-free (IC50 = 2.1 µM with FIDAS-5 vs. 4.9 µM with FIDAS-3; [L-Met] = [ATP] = 1 mM, [MAT2A] = 10 µg/mL; 20 min FIDAS preincubation prior to 30 min reaction) and cell-based (64% and 56% decrease of SAM and SAH level, respectively, in LS174T cells after 36 h treatment with either 3 µM FIDAS-5 or 10 µM FIDAS-3) assays. FIDAS-5 at 3 µM is shown to completely inhibit the proliferation of LS174T CRC (colorectal cancer) for up to 7 d in vitro and, when administered orally, effectively suppress HT29 CRC tumor expansion in mice in vivo (by 58% on d 18 post HT29 xenograft; 20 mg/kg/day starting d 4). Computer-aided in silico modeling predicts that FIDAS-3 & -5 target SAM-binding pocket at the interface of MAT2A dimer and in vitro binding studies reveals FIDAS-3 effectively competes against SAM for MAT2A binding.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MAT2A
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, W., et al. 2013. ACS Chem. Biol.8, 796.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service