Skip to Content
Merck
All Photos(2)

Key Documents

122335

Sigma-Aldrich

8-Quinolinol N-oxide

98%

Synonym(s):

8-Hydroxyquinoline N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$458.00

$458.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$458.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
128737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$458.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

135-138 °C (lit.)

SMILES string

Oc1cccc2ccc[n+]([O-])c12

InChI

1S/C9H7NO2/c11-8-5-1-3-7-4-2-6-10(12)9(7)8/h1-6,11H

InChI key

FJKUOCCQEBLPNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

8-Quinolinol N-oxide is a toxicant which can suppress luminescence of an indicator bacterium Photobacterium phosphoreum[1]. It forms two types of complexes with gold(III) and palladium(II): type I are ion-associated compounds containing hydrogen bond and the other is the six-membered metal chelates containing metal-oxygen bonds[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Sigler et al.
Folia microbiologica, 48(1), 95-102 (2003-05-15)
An easy-to construct, easy-to-operate standardized system was developed for determining the residual biological contamination of surgical instruments, endoscopes and other medical appliances subjected to hospital cleansing and/or disinfection. It consists of standard-sized pieces of glass, metal or endoscope plastic--dirt carriers--either
Syntheses of Gold (III) and Palladium (III) Complexes with 8-Quinolinol-N-Oxide and 5, 7-Dinitro-8-Quinolinol-N-Oxide.
Yuqiu G and Meichun J.
Precious Metals, 1, 002-002 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service