Skip to Content
Merck
All Photos(1)

Key Documents

494126

Sigma-Aldrich

Ghrelin, Human, Synthetic

The 28-amino acid n-octanoylated (at ser3) natural peptidyl ligand for the growth hormone secretagogue receptor 1a (GHSR1a).

Synonym(s):

Ghrelin, Human, Synthetic, H₂N-GS-S(n-octanoyl)-FLSPEHQRVQQRKESKKPPAKLQPR-CO₂H

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$317.00

$317.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$317.00

About This Item

Empirical Formula (Hill Notation):
C149H249N47O42
CAS Number:
Molecular Weight:
3370.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$317.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

The 28-amino acid n-octanoylated (at ser3) natural peptidyl ligand for the growth hormone secretagogue receptor 1a (GHSR1a). Ghrelin is secreted predominantly in the stomach and stimulates growth hormone release from the pituitary gland.

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Ser-Ser(n-Octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kojima, M. 2008. Regul. Pept.145, 2.
Wu, J.T., and Kral, J.G. 2004. Ann. Surg.239, 464
Kojima, M., et al. 1999. Nature402, 656.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service