Skip to Content
Merck
All Photos(1)

Key Documents

487870

Sigma-Aldrich

p-Nitrophenyl-β-D-xylopyranoside

Substrate for the colorimetric assay of β-D-xylosidase. Also shown to disrupt proteoglycan biosynthesis in vivo.

Synonym(s):

p-Nitrophenyl-β-D-xylopyranoside, pNP-β-D-Xyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$364.00

$364.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$364.00

About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$364.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% p-nitrophenol

solubility

water: 40 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)OC2OCC(C(C2O)O)O

InChI

1S/C11H13NO7/c13-8-5-18-11(10(15)9(8)14)19-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2

InChI key

MLJYKRYCCUGBBV-UHFFFAOYSA-N

General description

Substrate for the colorimetric assay of β-D-xylosidase. Also shown to disrupt proteoglycan biosynthesis in vivo.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Haggerty, J.G. 1994. J. Cell Physiol. 158, 39.
Gressner, A.M. 1991. Cell. Mol. Biol. 37, 549.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service