Skip to Content
Merck
All Photos(1)

Documents

48-660MAG

Millipore

MILLIPLEX MAPK/SAPK Signaling 10-Plex Kit - Cell Signaling Multiplex Assay

Synonym(s):

Cell Signaling Kit, MAPK/SAPK 10-Plex Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

assay range

inter-assay cv: 4-13%
intra-assay cv: 3-10%

detection method

fluorometric (Luminex xMAP)

storage temp.

2-8°C

General description

Cells respond to their environment in various ways through intracellular signaling. Signaling through Receptor Tyrosine kinases (RTKs) often promotes increased metabolism and cell growth. Activation of ERK/MAP kinase is one of the key serine/threonine kinases activated via RTK signaling, promoting increased activity of p70 S6 kinase, MSK1, STATs, CREB, and many other signaling intermediates. Other signaling pathways induced via stress (ex. heat shock/arsenite treatment) or death receptors (ex. TNFα) promote p38, JNK, and p53 signaling pathways.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • An overnight (4°C) incubation is recommended for best results.
  • This assay requires 25 μL diluted cell lysate.
  • This kit must be run using Assay Buffer 2 (provided).
  • 1 - 25 μg cell lysate/well (recommended starting concentration is 20 μg cell lysate/well).

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation or total pathway proteins in tissue and cell lysate samples.
Research Category
Signaling
Used to detect/quantify:ATF2 (Thr71)|Erk (Thr185/Tyr187)|HSP27 (Ser78)|JNK (Thr183/Tyr185)|c-Jun (Ser73)|MEK1 (Ser222)|MSK1 (Ser212)|p38 (Thr180/Tyr182)|p53 (Ser15)|STAT1 (Tyr701)

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Wei Wu et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 246-247, 110461-110461 (2020-06-05)
In arid conditions, the African Clawed frog Xenopus laevis enters a state of estivation dormancy as an adaptive survival strategy. Under estivation, X. laevis experience severe dehydration stress as 25-35% of total body water is lost. Dehydration in X. laevis

Related Content

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service