Skip to Content
Merck
All Photos(1)

Documents

48-614MAG

Millipore

MILLIPLEX® TGFβ Signaling Pathway Magnetic Bead 6-Plex - Cell Signaling Multiplex Assay

Synonym(s):

Cell Signaling Assay, Luminex® TGFβ Cell Signaling Assay, Millipore TGFβ Cell Signaling Assay, TGFβ Signal Transduction Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Cells respond to their environment in many different ways through intracellular signaling. The TGFβ pathway plays a central role in a number of normal cellular processes including proliferation, differentiation, apoptosis, migration, adhesion, immune response and other functions in most cells. Although the TGFβ pathway regulates a wide range of processes, the pathway is fairly simple. TGFβ dimers bind to TGFβ Type II Receptor which recruits and phosphorylates TGFβ Type I Receptor. The Type I Receptor then recruits and phosphorylates SMAD2/3, which then binds to SMAD4 and forms a complex that enters the cell nucleus where it acts as a transcription factor for various genes. TGFβ Receptor activates the SMAD-dependent canonical pathway, as well as SMAD-independent non-canonical pathways, such as PI3K/Akt, ERK, JNK, and p38. Aberrant TGFβ signaling is involved in the development of multiple diseases, including hematological malignancies such as leukemia, and impaired wound healing, neurodegenerative diseases, developmental disorders and pulmonary hypertension. Loss-of-function mutations promote tumorigenesis by suppressing the immune system and the epithelial mesenchymal transition (EMT), although TGFβ signaling has also been implicated in tumor inhibition. Immune cells such as B cells, T cells, dendritic cells and macrophages secrete TGFβ, which downregulates their proliferation, differentiation and activation by other cytokines. Consequently, modifications in TGFβ signaling have been linked to autoimmune and inflammatory diseases and cancer.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 1 (provided).1 - 25 μg cell lysate/well (recommended starting concentration is 40 to 1,000 μg protein/mL).Analytes available:Akt (Ser473);ERK (Thr185/Tyr187);Smad2 (Ser465/Ser467);Smad3 (Ser423/Ser425);Smad4 (Total);TGFβRII (Total)

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service