Skip to Content
Merck
All Photos(2)

Key Documents

482740

Sigma-Aldrich

nNOS - PSD-95 Interaction Inhibitor, ZL006

The nNOS - PSD-95 Interaction Inhibitor, ZL006 controls the biological activity of nNOS. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

nNOS - PSD-95 Interaction Inhibitor, ZL006, 5-(3,5-dichloro-2-hydroxybenzylamino)-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$281.00

$281.00


Estimated to ship on22 May 2025



Select a Size

Change View
10 MG
$281.00

About This Item

Empirical Formula (Hill Notation):
C14H11Cl2NO4
Molecular Weight:
328.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$281.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1c(c(cc(c1)Cl)CNc2cc(c(cc2)C(=O)O)O)O

InChI

1S/C14H11Cl2NO4/c15-8-3-7(13(19)11(16)4-8)6-17-9-1-2-10(14(20)21)12(18)5-9/h1-5,17-19H,6H2,(H,20,21)

InChI key

RTEYSQSXRFVKTJ-UHFFFAOYSA-N

General description

A cell-permeable and blood-brain barrier-permeant aminosalicylic acid compound that selectively disrupts nNOS and PSD-95 interaction upon NMDAR activation both in primary murine cortical neuron cultures (0.8- and 2.8-fold increase from basal level with or without 30 min 10 uM ZL006 pretreatment before 30 min Glu-Gly stimulation) in vitro and in mice subjected to middle brain artery occlusion (0.4- and 1.9-fold increase from basal level with or without inhibitor pretreatment 15 min before MACO; 1.5 mg/kg i.v.) in vivo without affecting nNOS activity or non-nNOS-mediated NMDAR EPSCs (excitatory postsynaptic currents) in rat hippocampus. Its therapeutic efficacy in limiting ischemia-caused brain tissue damage has been demonstrated in both mice and rats when applied 1 h after blood reperfusion following MACO (60% and 50% reduction of infarction, respectively; 1.5 mg/kg i.v.).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhou, L., et al. 2010. Nature Med.16, 1439.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service