Skip to Content
Merck
All Photos(1)

Key Documents

4810-OP

Millipore

Glycine

OmniPur® Grade, >= 98.5%, molecular biology grade

Synonym(s):

Glycine, Aminoacetic Acid, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$61.10
5 KG
$534.00

$61.10


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
$61.10
5 KG
$534.00

About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.28

$61.10


Estimated to ship on07 May 2025


Request a Bulk Order

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Suitable for a wide range of molecular biology applications. Molecular biology grade.

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected
Phosphatase: none detected
A260 (0.05 M, water): ≤0.07 AU
A280 (0.05 M, water): ≤0.07 AU
Purity: ≥98.5% by assay
Heavy metals (such as Pb): ≤0.001%






Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service