Skip to Content
Merck
All Photos(1)

Key Documents

475863

Sigma-Aldrich

MK2a Inhibitor

The MK2a Inhibitor, also referenced under CAS 41179-33-3, controls the biological activity of MK2a. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MK2a Inhibitor, Mitogen-activated protein kinase-activated protein kinase 2a Inhibitor, 4-(2ʹ-Fluorobiphenyl-4-yl)-N-(4-hydroxyphenyl)-butyramide, CMPD1, MAPKAPK2a Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$400.00

$400.00


Estimated to ship on05 May 2025



Select a Size

Change View
5 MG
$400.00

About This Item

Empirical Formula (Hill Notation):
C22H20FNO2
CAS Number:
Molecular Weight:
349.40
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$400.00


Estimated to ship on05 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(cccc1)c2ccc(cc2)CCCC(=O)Nc3ccc(cc3)O

InChI

1S/C22H20FNO2/c23-21-6-2-1-5-20(21)17-10-8-16(9-11-17)4-3-7-22(26)24-18-12-14-19(25)15-13-18/h1-2,5-6,8-15,25H,3-4,7H2,(H,24,26)

InChI key

ODYAQBDIXCVKAE-UHFFFAOYSA-N

General description

A non-ATP-competitive, reversible, selective inhibitor of p38α-mediated MK2a (mitogen-activated protein kinase-activated protein kinase 2a) phosphorylation (Kiapp = 330 nM). Does not inhibit p38α-mediated phosphorylation of the the other two known p38 substrates, MBP and ATF-2.
A reversible p-amidophenolic compound that selectively inhibits the phosphorylation of MK2a (mitogen-activated protein kinase-activated protein kinase 2a; Kiapp = 330 nM) by p38α in a non-ATP-competitive manner. Does not block the kinase activity of p38α towards the other two known p38 substrates, MBP and ATF-2.

Biochem/physiol Actions

Cell permeable: no
Kiapp = 330 nM against mitogen-activated protein kinase-activated protein kinase 2a (MK2a)
Primary Target
Mk2α
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Davidson, W., et al. 2004. Biochemistry43, 11658.
Lukas, S.M., et al. 2004. Biochemistry43, 9950.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service