Skip to Content
Merck
All Photos(1)

Key Documents

400012

Sigma-Aldrich

Caspase-1 Inhibitor II

The Caspase-1 Inhibitor II, also referenced under CAS 178603-78-6, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-1 Inhibitor II, IL-1β Converting Enzyme (ICE) Inhibitor II, Ac-YVAD-CMK

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$525.00

$525.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 MG
$525.00

About This Item

Empirical Formula (Hill Notation):
C24H33ClN4O8
CAS Number:
Molecular Weight:
540.99
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

$525.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

ClCC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)Cc1ccc(cc1)O)C(C)C)C)CC(=O)O

InChI

1S/C24H33ClN4O8/c1-12(2)21(24(37)26-13(3)22(35)28-17(10-20(33)34)19(32)11-25)29-23(36)18(27-14(4)30)9-15-5-7-16(31)8-6-15/h5-8,12-13,17-18,21,31H,9-11H2,1-4H3,(H,26,37)(H,27,30)(H,28,35)(H,29,36)(H,33,34)/t13-,17-,18-,21-/m0/s1

InChI key

UOUBHJRCKHLGFB-DGJUNBOTSA-N

General description

A cell-permeable and irreversible inhibitor of caspase-1 (Ki = 760 pM), caspase-4, and caspase-5. Inhibits Fas-mediated apoptosis and acidic sphingomyelinase activation.
Interleukin-1β converting enzyme (ICE) is a cysteine protease that cleaves the IL-1β precursor to mature IL-1β. Caspase-1 inhibitor II is a cell-permeable, irreversible inhibitor of Caspase-1, Caspase-4, and Caspase-5. Inhibits CPP32/Apopain at a much higher concentration. Inhibits Fas-mediated apoptosis and acidic sphingomyelinase activation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1
Product does not compete with ATP.
Reversible: no
Target Ki: 760 pM against caspase-1

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Tyr-Val-Ala-Asp-CMK

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Brenner, B., et al. 1998. Cell Death Differ. 5, 29.
Garcia-Calvo, M., et al. 1998. J. Biol. Chem. 273, 32608.
Hilbi, H., et al. 1998. J. Biol. Chem. 273, 32895.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Schlegel, J., et al. 1996. J. Biol. Chem.271, 1841.
Enari, M., et al. 1995. Nature375, 78.
Walker, N.P.C., et al. 1994. Cell78, 343.
Thornberry, N., et al. 1992. Nature356, 768.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bohm Lee et al.
The Journal of biological chemistry, 298(3), 101647-101647 (2022-02-02)
The dual leucine zipper kinase (DLK) is a key regulator of axon regeneration and degeneration in response to neuronal injury; however, regulatory mechanisms of the DLK function via its interacting proteins are largely unknown. To better understand the molecular mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service