Skip to Content
Merck
All Photos(2)

Key Documents

422689

Sigma-Aldrich

Kir2.1 Inhibitor, ML133

The Kir2.1 Inhibitor, ML133, also referenced under CAS 185669-79-8, controls the biological activity of Kir2.1. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Kir2.1 Inhibitor, ML133, Inward rectifier potassium channel blocker, ML133

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$283.00

$283.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
$283.00

About This Item

Empirical Formula (Hill Notation):
C19H19NO
CAS Number:
Molecular Weight:
277.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$283.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

COC(C=C1)=CC=C1CNCC2=C(C=CC=C3)C3=CC=C2

InChI

1S/C19H19NO/c1-21-18-11-9-15(10-12-18)13-20-14-17-7-4-6-16-5-2-3-8-19(16)17/h2-12,20H,13-14H2,1H3

InChI key

GIFYKGIODMXYNT-UHFFFAOYSA-N

General description

A cell-permeable, renal outer medullary potassium (ROMK) blocker that is shown to inhibit inward rectifier potassium channel, Kir2.1 (IC50 = 1.8 µM and 290 nM, at pH 7.4 and pH 8.5, respectively). It exhibits little selectivity against other members of the Kir2.x family, but has no effect on Kir1.1 (IC50 >300 µM) and displays weak activity for Kir4.1 and Kir7.1 (IC50 = 76 µM and 33 µM, respectively). It is reported to be assayed in 218 MLPCN HTS campaigns, demonstrating activity in only two assays that are not based on Kir2.1. Furthermore, it displays a relatively clean profile in a radioligand binding panel of 68 GPCRs, ion channels and transporters.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lindsley, C.W., et al. 2011. ACS Chem. Biol.6, 845.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polarized localization of phosphatidylserine in the endothelium regulates Kir2.1.
Ruddiman, et al.
JCI insight, 8 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service