420411
Kanamycin Sulfate, Streptomyces kanamyceticus, Cell Culture-Tested
An aminoglycoside antibiotic effective against Gram-positive and Gram-negative organisms. Inhibitor of protein biosynthesis that acts on the 30S ribosome, causing misreading of the genetic code.
Synonym(s):
Kanamycin Sulfate, Streptomyces kanamyceticus, Cell Culture-Tested
About This Item
Recommended Products
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white to off-white
solubility
water: 10 mg/mL
shipped in
ambient
storage temp.
10-30°C
InChI
1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1
InChI key
OOYGSFOGFJDDHP-KMCOLRRFSA-N
General description
Biochem/physiol Actions
protein biosynthesis that acts on the 30S ribosome
Warning
Reconstitution
Other Notes
Cardon, G.H., et al. 1993. Plant J.3, 773.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition. Cold Spring Harbor, New York. p.1.6, A.6.
Korzybyski, T., et al. 1978. In Antibiotics, Origin, Nature, and Properties1, 690.
Stiffler, P.W., et al. 1974. Antimicrob. Agents Chemother.6, 516.
Umezawa, S., et al. 1968. J. Antibiot.21, 367.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service