Skip to Content
Merck
All Photos(2)

Key Documents

G1914

Sigma-Aldrich

Gentamicin sulfate

meets USP testing specifications, powder

Synonym(s):

Gentamicin sulfate salt, Garamycin, Gentiomycin C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51281651
eCl@ss:
42020823
PubChem Substance ID:
NACRES:
NA.76

biological source

Micromonospora purpurea

Quality Level

Agency

USP/NF
meets USP testing specifications

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

application(s)

pharmaceutical (small molecule)

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O=S(O)(O)=O.O[C@]1(C)[C@@H]([C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]3O[C@@](CC[C@H]3N)([C@@H](C)NC)[H])[C@@H](N)C[C@H]2N)OC1)NC.O[C@]4(C)[C@@H]([C@@H](O)[C@@H](O[C@@H]5[C@@H](O)[C@H](O[C@H]6O[C@@](CC[C@H]6N)([C@@H](C)N)[H])[C@@H](N)C[C@H]5N)OC4)NC.O[

InChI

1S/C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;1-8(21)12-5-4-9(22)18(30-12)31-15-10(23)6-11(24)16(13(15)26)32-19-14(27)17(25-3)20(2,28)7-29-19;1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;1-5(2,3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;8-19,25-28H,4-7,21-24H2,1-3H3;8-18,24-27H,3-7,20-23H2,1-2H3;(H2,1,2,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;8-,9-,10+,11-,12+,13+,14-,15-,16+,17-,18-,19-,20+;8-,9+,10-,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m110./s1

InChI key

RDEIXVOBVLKYNT-HDZPSJEVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Gentamicin sulfate, a broad-spectrum antibiotic, used as a selection agent (gentamicin-resistance gene) in cell culture and molecular biology applications. This product is recommended for use at 50 mg/L.

Biochem/physiol Actions

Mode of action: Gentamicin causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site. The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilizes the membrane, and forms holes in the cell surface.

Antimicrobial spectrum: Includes Gram-negative and Gram-positive bacteria, including strains resistant to tetracycline, chloramphenicol, kanamycin and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus and Streptococcus.

Components

Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt. There are three components, each consisting of five basic nitrogens and requiresing five equivalents of sulfuric acid per mole of gentamicin base.

Caution

Sterile solutions should be stored at 2-8°C. Solutions have also been shown to be stable at room temperature and in boiling aqueous buffers of pH 2-142.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erika E A Brown et al.
PloS one, 11(11), e0166989-e0166989 (2016-11-22)
The morphology and distribution of lateral line neuromasts vary between ecomorphological types of anuran tadpoles, but little is known about how this structural variability contributes to differences in lateral-line mediated behaviors. Previous research identified distinct differences in one such behavior
Chang-Chin Wu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(1), 80-87 (2016-11-20)
Patients with inadequate volume of alveolar processes or bone defects commonly require graft substitutes in oral, maxillofacial or orthopedic surgery. Ridge augmentation and reconstruction of facial bony defects with bone graft materials achieve better outcomes in functional and aesthetic rehabilitation.
Artur Adamczak et al.
Journal of clinical medicine, 9(1) (2020-01-08)
Among natural substances widespread in fruits, vegetables, spices, and medicinal plants, flavonoids and organic acids belong to the promising groups of bioactive compounds with strong antioxidant and anti-inflammatory properties. The aim of the present work was to evaluate the antibacterial
Changjun Han et al.
Biomacromolecules, 18(11), 3776-3787 (2017-10-05)
In addition to customizing shapes of metal bone substitutes for patients, the 3D printing technique can reduce the modulus of the substitutes through the design and manufacture of interconnected porous structures, achieving the modulus match between substitute and surrounding bone
David Ochoa et al.
Molecular systems biology, 12(12), 888-888 (2016-12-03)
The coordinated regulation of protein kinases is a rapid mechanism that integrates diverse cues and swiftly determines appropriate cellular responses. However, our understanding of cellular decision-making has been limited by the small number of simultaneously monitored phospho-regulatory events. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service