Skip to Content
Merck
All Photos(2)

Key Documents

420351

Sigma-Aldrich

ICMT Inhibitor II, FTPAT

The ICMT Inhibitor II, FTPAT controls the biological activity of ICMT.

Synonym(s):

ICMT Inhibitor II, FTPAT, ( E)-methyl-3-((5-(4-(2-((1,1ʹ-biphenyl]-4-yl)ethyl)-1H-1,2,3-triazol-1-yl)-3-methylpent-2-en-1-yl)thio)propanoate, (E)-methyl-3-((5-(4-(2-((1,1ʹ-biphenyl]-4-yl)ethyl)-1H-1,2,3-triazol-1-yl)-3-methylpent-2-en-1-yl)thio)propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$342.00

$342.00


Estimated to ship on23 April 2025



Select a Size

Change View
10 MG
$342.00

About This Item

Empirical Formula (Hill Notation):
C26H31N3O2S
CAS Number:
Molecular Weight:
449.61
UNSPSC Code:
12352200
NACRES:
NA.77

$342.00


Estimated to ship on23 April 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(OC)CCSC/C=C(C)/CCN1N=NC(CCC2=CC=C(C3=CC=CC=C3)C=C2)=C1

InChI key

LUXQNURJPSSXSC-RCCKNPSSSA-N

General description

A cell-permeable triazolo-farnesylthiopropanoate compound that acts as a potent, reversible, and isoprenyl mimetic competitive inhibitor of isoprenylcysteine carboxyl methyltransferase activity (ICMT; IC50 = 800 nM; Ki = 400 nM). Shown to prevent K-Ras membrane localization in GFP-K-Ras transfected Jurkat T-cells. Exhibits selective toxicity towards wt-Icmt+/+ mouse embryonic fibroblasts (MEFs) over Icmt-/- MEFs (IC50 = 33 and >100 µM). Also shown to arrest the proliferation of PaTu-8902, a highly metastatic pancreatic cancer cells (IC50 = 8 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bergman, J.A., et al. 2012. ACS Med. Chem. Lett.3, 15.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service