Skip to Content
Merck
All Photos(1)

Documents

420350

Sigma-Aldrich

ICMT Inhibitor

The ICMT Inhibitor controls the biological activity of ICMT. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

ICMT Inhibitor, 2-(5-(3-Methylphenyl)-1-octyl-1H-indol-3-yl)acetamide, 2-(1-Octyl-5-m-tolyl-1H-indol-3-yl) acetamide, Cysmethynil, Cysmethynil, Isoprenylcysteine Carboxyl Methyltransferase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32N20
Molecular Weight:
612.66
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable indole acetamide compound that inhibits Icmt (isoprenylcysteine carboxyl methyltransferase) activity in an acceptor substrate isoprenylated cysteine-competitive (Ki = 0.02 µM; IC50 = 0.29 and 2.1 µM, respectively, with or without inhibitor preincubation; Substrate = S-farnesyl-L-cysteine) and donor substrate AdoMet-noncompetitive manner, exhibiting no activity against FTase, gernaylgeranyltransferase type I, CaaX protease Rce1, AdoMet-dependent DNA methyltransferase, SssI DNA methyltransferase, or PCMT1 protein methyltransferase. Shown to inhibit the proliferation of non-cancer MEF cells (by 100% at 30 µM) and prevent anchorage-independent growth of colon cancer DKOB8 (by >90% at 20 µM) in a dose- and Icmt-dependent manner. Cysmethynil (25 µM) induces G1 cell cycle-arrest and autophagy-mediated, but not apoptotic, cell death in prostate cancer PC3 cells with concomitant PI 3-K/Akt and mTOR signaling blockage, consistent with inhibition of Icmt-mediated activation of Ras and Rheb GTPases. Murine PC3 xenograft studies reveal limited in vivo efficacy (tumor volume on day 28 = 9- and 22-fold of day 0, respectively, with or without 200 mg/kg/i.p. q.o.d.).
A cell-permeable indole acetamide that acts as a substrate-competitive and AdoMet-noncompetitive Icmt (isoprenylcysteine carboxyl methyltransferase) inhibitor (Ki = 0.02 µM; IC50 = 0.29 µM; Substrate = S-farnesyl-L-cysteine), exhibiting no activity against FTase, gernaylgeranyltransferase type I, CaaX protease Rce1, AdoMet-dependent DNA methyltransferase, SssI DNA methyltransferase, or PCMT1 protein methyltransferase. Cysmethynil (25 µM) induces G1 cell cycle-arrest and autophagy-mediated, but not apoptotic, cell death in prostate cancer PC3 cells with concomitant PI 3-K/Akt and mTOR signaling blockage, consistent with inhibition of Icmt-mediated activation of Ras and Rheb GTPases. Murine PC3 xenograft studies reveal limited in vivo efficacy.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C).

Other Notes

Go, M.L., et al. 2010. J. Med. Chem.53, 6838; Wang, M., et al. 2008. J. Biol. Chem.283, 18678; Baron, R.A., et al. 2007. Biochemistry46, 554; Winter-Vann, A.M., et al. 2005. Proc. Natl. Acad. Sci. USA102, 4336.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service