Skip to Content
Merck
All Photos(1)

Key Documents

420294

Sigma-Aldrich

STAT5 Inhibitor II, IQDMA

The STAT5 Inhibitor II, IQDMA controls the biological activity of STAT5.

Synonym(s):

STAT5 Inhibitor II, IQDMA, N1-(11H-indolo[3,2-c]quinolin-6-yl)-N2,N2-dimethylethane-1,2-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$210.00

$210.00


Estimated to ship on01 June 2025



Select a Size

Change View
10 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C19H20N4
Molecular Weight:
304.39
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable indoloquinoline compound that exhibits anticancer activity and inhibits the growth of A549 lung cancer cells (IC50 = 8 µM), as well as HL-60 (IC50 = 5 µM) and K562 (IC50 = 8 µM) leukemia cells. IQDMA-mediated apoptosis is shown to correlate with a downregulation of STAT5 pathway signaling in HL-60 and K562 cultures, while JNK and p38 signalings are shown to be activated in A549 cells upon IQDMA treatment.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Su, J.C., et al. 2009, Life Sci.85, 505.
Chien, C.M., et al. 2008, Chem-Biol. Intr.176, 40.
Yang, S.H., et al. 2008, J. Biochem. Mol. Tox.22, 396.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Castillo et al.
American journal of cancer research, 12(1), 396-426 (2022-02-11)
Pregnancy-associated breast cancer (PABC) is diagnosed during or shortly after pregnancy. Although rare, PABC is a serious occurrence often of the triple negative (TNBC) subtype. Here we show progesterone, prolactin, and RANKL upregulate BRCA1-IRIS (IRIS) in separate and overlapping subpopulations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service