Skip to Content
Merck
All Photos(1)

Key Documents

407951

Millipore

Ionomycin, Free Acid, Streptomyces conglobatus in Solution

Synonym(s):

Ionomycin, Free Acid, Streptomyces conglobatus in Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$286.00

$286.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$286.00

About This Item

Empirical Formula (Hill Notation):
C41H72O9
CAS Number:
Molecular Weight:
709.00
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.76

$286.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (HPLC)

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

clear colorless to slight yellow

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C41H72O9/c1-25(21-29(5)34(43)24-35(44)30(6)22-27(3)20-26(2)14-15-38(46)47)12-11-13-28(4)39(48)31(7)36(45)23-33-16-18-41(10,49-33)37-17-19-40(9,50-37)32(8)42/h11,13,24-33,36-37,39,42-43,45,48H,12,14-23H2,1-10H3,(H,46,47)/b13-11+,34-24-/t25-,26-,27+,28-,29-,30+,31+,32-,33?,36+,37?,39-,40+,41+/m1/s1

InChI key

PGHMRUGBZOYCAA-OJFQOPKISA-N

General description

Highly specific for divalent cations (Ca2+ > Mg2+ >> Sr2+ = Ba2+). More effective than antibiotic A23187 as a mobile ion carrier for Ca2+. Complexes with Ca2+ between pH 7 and 9.5, causing significant absorption in the UV range; however, in contrast to A23187 (Cat. No. 100105), it is not fluorescent. Useful for studies of Ca2+ transport across biological membranes and measurement of cytoplasmic free Ca2+. Induces apoptotic neuronal degeneration in embryonic cortical neurons. Also induces cell cycle arrest in the G1 interval of mature Burkitt lymphoma cell lines.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Mobile ion carrier for Ca2+.
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 5 mM (1 mg/282 µl) solution of Ionomycin, Free Acid, Streptomyces conglobatus (Cat. No. 407950) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Aagaard-Tillery, K.M., and Jelinek, D.F. 1995. J. Immunol.155, 3297.
Takei, N., and Endo, Y. 1994. Brain Res. 652, 65.
Dupuy d’Angeac, A., et al. 1993. J. Cell Physiol.154, 44.
Lee, H.C. 1993. J. Biol. Chem.268, 293.
Kuznetsov, G., et al. 1992. J. Biol. Chem. 267, 3932.
Toeplitz, B.K., et al. 1979. J. Am. Chem. Soc.101, 3344.
Liu, C.M., and Hermann, T.E. 1978. J. Biol. Chem.253, 5892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beibei Wu et al.
Cell reports, 36(8), 109603-109603 (2021-08-26)
Sustained activation of the Ca2+-release-activated Ca2+ (CRAC) channel is pivotal for effector T cell responses. The mechanisms underlying this sustainability remain poorly understood. We find that plasma membrane localization of ORAI1, the pore subunit of CRAC channels, is limited in effector
Fang-Yuan Mao et al.
Cellular and molecular gastroenterology and hepatology, 12(2), 395-425 (2021-03-07)
Rev-erbα represents a powerful transcriptional repressor involved in immunity. However, the regulation, function, and clinical relevance of Rev-erbα in Helicobacter pylori infection are presently unknown. Rev-erbα was examined in gastric samples from H pylori-infected patients and mice. Gastric epithelial cells (GECs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service