Skip to Content
Merck
All Photos(1)

Key Documents

407653

Sigma-Aldrich

Interleukin-6 Receptor, Soluble, Human, Recombinant, S. frugiperda

Synonym(s):

rhIL-6 sR

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$1,630.00

$1,630.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 μG
$1,630.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.42

$1,630.00


Estimated to ship on28 May 2025


Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human soluble interleukin-6 receptor expressed in S. frugiperda insect cells. Soluble form of the IL-6 receptor that binds IL-6 and mediates IL-6 signaling through interaction with gp 130. Elevated serum levels of soluble IL-6 receptors have been shown to be associated with a number of pathological states, including multiple myelomas, adult T cell leukemia, interstitial lung infection, and HIV infection.

Biochem/physiol Actions

ED₅₀= ≥5 ng/ml as measured by its ability to enhance IL-6-induced growth inhibition of mouse M1 myeloid leukemic cells

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS, 50 µg BSA/µg IL-6 receptor.

Reconstitution

Reconstitute to a concentration ≥100 µg/ml with sterile PBS containing at least 0.1% HSA or BSA.

Other Notes

Schobitz, B., et al. 1995. FASEB J. 9, 659.
Yokoyama, A., et al. 1995. Clin. Exp. Immunol. 100, 325.
Suzuki, H., et al. 1993. Eur. J. Immunol. 23, 1078.
Lust, J.A., et al. 1992. Cytokine 4, 96.
Sugita, T., et al. 1990. J. Exp. Med. 171, 2001.
Yamasaki, K., et al. 1988. Science241, 825.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service