Skip to Content
Merck
All Photos(1)

Documents

407290

Sigma-Aldrich

Anti-α-Interferon Mouse mAb (MMHA-2)

liquid, clone MMHA-2, Calbiochem®

Synonym(s):

Anti-IFN-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MMHA-2, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNA1(3439)

General description

Purified mouse monoclonal antibody. Recognizes the ~19 kDa human α-interferon (KA ≈109 M-1).
Recognizes the ~19 kDa IFN-α protein.
This Anti-α-Interferon Mouse mAb (MMHA-2) is validated for use in ELISA, Immunoblotting, IHC, IP, Neutralization Studies for the detection of α-Interferon.

Immunogen

α-interferon from human leukocytes

Application

ELISA (see comments)

Immunoblotting (1:500)

Immunohistochemistry (see comments)

Immunoprecipitation (see comments)

Neutralization Studies (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 18 mM Citric Acid, 165 mM Sodium Phosphate Dibasic.

Other Notes

Familletti, P.C., et al. 1981. Methods Enzymol. 78, 387.
Rubinstein, S., et al. 1981. J. Virol. 37, 755.
Suitable for neutralization of human α-interferon. Does not recognize β-interferon or γ-interferon by immunoblotting. Effectively neutralizes human α-interferon. This antibody has also been reported to work with ELISA, immunohistochemistry, and immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service