Skip to Content
Merck
All Photos(1)

Key Documents

401482

Sigma-Aldrich

IKK-2 Inhibitor V

The IKK-2 Inhibitor V, also referenced under CAS 978-62-1, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IKK-2 Inhibitor V, N-(3,5-Bis-trifluoromethylphenyl)-5-chloro-2-hydroxybenzamide, IMD-0354

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$540.00

$540.00


Estimated to ship on17 April 2025



Select a Size

Change View
5 MG
$540.00

About This Item

Empirical Formula (Hill Notation):
C15H8ClF6NO2
CAS Number:
Molecular Weight:
383.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$540.00


Estimated to ship on17 April 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)NC(=O)c2c(ccc(c2)Cl)O

InChI

1S/C15H8ClF6NO2/c16-9-1-2-12(24)11(6-9)13(25)23-10-4-7(14(17,18)19)3-8(5-10)15(20,21)22/h1-6,24H,(H,23,25)

InChI key

CHILCFMQWMQVAL-UHFFFAOYSA-N

General description

A cell-permeable IKK-2 inhibitor that selectively blocks IκBα phosphorylation (IC50 ~250 nM) and prevents the induction of NF-κB p65 nuclear translocation. Reported to suppress neoplastic proliferation of mast cells with constitutively activated c-kit receptor. Shown to offer cardioprotection by reducing IL-1β and MCP-1 production (IC50<1 µM) in cardiomyocytes, and ameliorate insulin resistance in diabetic KKAy mice by regulating adiponectin release.
A cell-permeable salicylamide compound that acts as an IKK-2 inhibitor by selectively blocking IκBα phosphorylation (IC50 ~ 250 nM) and thereby prevents the induction of NF-κB p65 nuclear translocation. Shown to offer cardioprotection by reducing IL-1β and MCP-1 production (IC50<1 µM) in cardiomyocytes, and ameliorate insulin resistance in KKAy mice by regulating adiponectin release.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IKK-2
Product does not compete with ATP.
Reversible: no
Target IC50: ~ 250 nM blocking IκBα phosphorylation

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kamon, J., et al. 2004. Biochem. Biophys. Res. Commun.323, 242.
Onai, Y., et al. 2004. Cardiovasc. Res.63, 51.
Tanaka, A., et al. 2004. Blood105, 2324.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service