394111
BIS-TRIS propane
ULTROL® Grade, >98% (titration), non-zwitterionic buffer useful in pH range 6.3-9.5
Synonym(s):
BIS-Tris Propane, ULTROL® Grade, 1,3- bis[ tris(Hydroxymethyl)methylamino]propane, 1,3-bis[tris(Hydroxymethyl)methylamino]propane
Select a Size
About This Item
Recommended Products
Quality Level
Assay
>98% (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
pKa (20 °C)
6.8
solubility
water: 1 M
cation traces
heavy metals: <1 ppm
shipped in
ambient
storage temp.
15-25°C
SMILES string
N(C(CO)(CO)CO)CCCNC(CO)(CO)CO
InChI
1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2
InChI key
HHKZCCWKTZRCCL-UHFFFAOYSA-N
Related Categories
General description
Application
- Comparing solution-gate and bottom-gate nanowire field-effect transistors on pH sensing with different salt concentrations and surface modifications.: BIS-TRIS propane is used as a buffering agent in the development of nanowire field-effect transistors for pH sensing. The research demonstrates its effectiveness in enhancing the sensitivity and stability of these biosensors under varying conditions (Hu et al., 2024).
- Laser-induced tuning of graphene field-effect transistors for pH sensing.: This study employs BIS-TRIS propane as a buffering solution to optimize the performance of graphene field-effect transistors in pH sensing applications. The findings highlight the role of BIS-TRIS propane in achieving precise tuning of sensor responses (Lampinen et al., 2023).
- Elucidating the exchange interactions in a {Gd(III)Cu(II)(4)} propellor.: BIS-TRIS propane is utilized as a buffer in the investigation of exchange interactions in complex metal assemblies. The study provides insights into the magnetic properties and potential applications in materials science (Heras Ojea et al., 2023).
- Protein association on multimodal chromatography media.: This research explores the use of BIS-TRIS propane in multimodal chromatography to study protein interactions. The buffer′s compatibility with various chromatographic techniques enhances the understanding of protein behavior and purification processes (Muca & Antos, 2023).
Warning
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service