Skip to Content
Merck
All Photos(1)

Key Documents

391335

Millipore

BIS-TRIS

ULTROL® Grade, ≥99% dry basis (titration), suitable where a calcium phosphate-DNA complex formation is desirable

Synonym(s):

BIS-Tris, ULTROL® Grade, [ bis(2-Hydroxyethyl)imino]- tris(hydroxymethyl)methane, [bis(2-Hydroxyethyl)imino]-tris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$277.00
1 KG
$1,340.00

$277.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$277.00
1 KG
$1,340.00

About This Item

Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

$277.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

6.5

solubility

water: soluble

cation traces

heavy metals: <3 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

N(C(CO)(CO)CO)(CCO)CCO

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2

InChI key

OWMVSZAMULFTJU-UHFFFAOYSA-N

General description

A zwitterionic buffer that exhibits only a very small shift in dissociation constant with respect to temperature. Useful in the pH 5.8-7.3 range. Has a pKa of 6.50 at 25°C. Suitable for fractionation of hemoglobin.
Absorbance (100 mM, H2O, 280 nm):≤0.04.
An inert, zwitterionic buffer that exhibits only a very small shift in dissociation constant with respect to temperature (absorbance: <0.04 at 280 nm, 100 mM, H2O). Suitable for fractionation of hemoglobin. Has a pKa of 6.5 at 25°C.

Warning

Toxicity: Standard Handling (A)

Other Notes

Nealon, D.A., et al. 1980. Clin Chem.26, 1516.
Morin, L.G. 1977. Clin. Chem.23, 1569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service