Skip to Content
Merck
All Photos(2)

Key Documents

385570

Sigma-Aldrich

Heat Shock Factor 1 Inhibitor, KRIBB11

The Heat Shock Factor 1 Inhibitor, KRIBB11 controls the biological activity of Heat Shock Factor 1.

Synonym(s):

Heat Shock Factor 1 Inhibitor, KRIBB11, Hsp70 Induction Inhibitor II, N²-(1H-Indazol-5-yl)-N⁶-methyl-3-nitropyridine-2,6-diamine, HSF1 Inhibitor, KRIBB11, Heat Shock Protein Inhibitor III, p-TEFb hsp70 Promoter Recruitment Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N6O2
CAS Number:
Molecular Weight:
284.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-yellow

solubility

DMSO: 50 mg/mL, brown

shipped in

ambient

storage temp.

−20°C

SMILES string

CNC(C=C1)=NC(NC2=CC3=C(NN=C3)C=C2)=C1[N+]([O-])=O

InChI

1S/C13H12N6O2/c1-14-12-5-4-11(19(20)21)13(17-12)16-9-2-3-10-8(6-9)7-15-18-10/h2-7H,1H3,(H,15,18)(H2,14,16,17)

InChI key

NDJJEQIMIJJCLL-UHFFFAOYSA-N

General description

A cell-permeable 2,6-diaminopyridine compound that interacts with HSF1 (heat shock factor 1) in a reversible manner and prevents HSF1 from recruiting p-TEFb (positive transcription elongation factor b) to the hsp70 promoter site (IC50 ≤3 µM in HCT-116 ChIP assays), a critical step for the p-TEFb CDK9 subunit-catalyzed pol II CTD (RNA polymerase II c-terminal domain) Ser-2 phosphorylation and transcription activation upon heat shock induction (IC50<1, <3, <5, and <10 µM, respectively, against reporter, HSP47, HSP27, and HSP70 transcription in HCT-116 cultures). Reported to exhibit antiproliferation activity against several cancer cultures in vitro (IC50 ranges from 3 to 8 µM) and effectively suppress HCT-116-derived tumor growth in mice in vivo (by 47.4% on day 31; single i.p. dosage of 50 mg/kg on day 13 after cancer implantation). Does not affect TNF-α-induced NF-κB activity in HeLa or heat shock-induced HSF1 ser230 phosphorylation and promoter association in HCT-116 even at concentrations as high as 10 µM.
A cell-permeable 2,6-diaminopyridine compound that interacts with HSF1 in a reversible manner and prevents HSF1 from recruiting p-TEFb to the hsp70 promoter site (IC50 ≤3 µM in HCT-116 ChIP assays), a critical step for heat shock-induced transcription activation (IC50<1, <3, <5, and <10 µM, respectively, against reporter, HSP47, HSP27, and HSP70 transcription in HCT-116 cultures). Reported to exhibit antiproliferation activity against several cancer cultures in vitro (IC50 from 3 to 8 µM) and effectively suppress HCT-116-derived tumor growth in mice in vivo (50 mg/kg i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yoon, Y.J., et al. 2010. J. Biol. Chem.286, 1737.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Csaba András Schvarcz et al.
Cancers, 13(7) (2021-05-01)
Modulated electro-hyperthermia (mEHT) is a selective cancer treatment used in human oncology complementing other therapies. During mEHT, a focused electromagnetic field (EMF) is generated within the tumor inducing cell death by thermal and nonthermal effects. Here we investigated molecular changes
Young Ju Yoon et al.
The Journal of biological chemistry, 286(3), 1737-1747 (2010-11-17)
Heat shock factor 1 (HSF1) is the master switch for heat shock protein (HSP) expression in eukaryotes. A synthetic chemical library was screened to identify inhibitors of HSF1 using a luciferase reporter under the control of a heat shock element.
Lea Danics et al.
Cancers, 12(9) (2020-09-16)
Modulated electro-hyperthermia (mEHT) is a complementary antitumor therapy applying capacitive radiofrequency at 13.56 MHz. Here we tested the efficiency of mEHT treatment in a BALB/c mouse isograft model using the firefly luciferase-transfected triple-negative breast cancer cell line, 4T1. Tumors inoculated
Maruhen Ad Silveira et al.
Life science alliance, 4(5) (2021-02-18)
Master transcription factors control the transcriptional program and are essential to maintain cellular functions. Among them, steroid nuclear receptors, such as the estrogen receptor α (ERα), are central to the etiology of hormone-dependent cancers which are accordingly treated with corresponding
Greg A Timblin et al.
Nature metabolism, 3(5), 618-635 (2021-05-26)
Macrophages generate mitochondrial reactive oxygen species and mitochondrial reactive electrophilic species as antimicrobials during Toll-like receptor (TLR)-dependent inflammatory responses. Whether mitochondrial stress caused by these molecules impacts macrophage function is unknown. Here, we demonstrate that both pharmacologically driven and lipopolysaccharide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service