Skip to Content
Merck
All Photos(1)

Documents

345749

Sigma-Aldrich

GDF-8 Propeptide

≥90% (SDS-PAGE), lyophilized powder, mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.26

product name

GDF-8 Propeptide, His•Tag®, Mouse, Recombinant, GDF-8 Propeptide, His•Tag, Mouse, Recombinant, consists of amino acids 25-285, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-derived mouse myeloma cells.

recombinant

expressed in mouse NSO cells (myeloma cells)

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse growth differentiation factor-8 (GDF-8)/myostatin propeptide, consisting of amino acids 25-285, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-derived mouse myeloma cells. GDF-8 is a secreted protein that is expressed during embryonic development and in adult skeletal muscle.

Application

GDF-8 Propeptide, His•Tag, Mouse, Recombinant, consists of amino acids 25-285, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-derived mouse myeloma cells.

Biochem/physiol Actions

ED₅₀ = 0.05-0.25 µg/ml (using 40 ng/ml rmGDF-8) as measured by inhibition of rmGDF-8 activity in K562 human chronic myelogenous leukemia cells. Approximately 1 µg/ml will completely inhibit GDF-8 activity in K562 cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered PBS, BSA.

Reconstitution

Following reconstituyion refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 50 µg/ml with sterile PBS containing ≥1% human or bovine serum albumin.

Other Notes

Hill, J.J. et al. 2003. Molecular Endocrinology17, 1144.
Thies, R.S. et al. 2001. Growth Factors18, 251.
Chang, H. et al. 2002. Endocrine Reviews23, 787.
McPherron, A.C. et al. 1997. Nature387, 83.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service