Skip to Content
Merck
All Photos(3)

Key Documents

245917

Sigma-Aldrich

Pentafluoropropionic acid

97%

Synonym(s):

Perfluoropropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$138.00
50 G
$537.00

$138.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$138.00
50 G
$537.00

About This Item

Linear Formula:
CF3CF2COOH
CAS Number:
Molecular Weight:
164.03
Beilstein:
1773387
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$138.00


Available to ship on22 April 2025Details


Request a Bulk Order

vapor density

~5.6 (vs air)

Quality Level

vapor pressure

~40 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.284 (lit.)

bp

96-97 °C (lit.)

density

1.561 g/mL at 25 °C (lit.)

application(s)

PFAS testing

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)C(F)(F)C(F)(F)F

InChI

1S/C3HF5O2/c4-2(5,1(9)10)3(6,7)8/h(H,9,10)

InChI key

LRMSQVBRUNSOJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The kinetics of the reaction of pentafluoropropionic acid with fluorine atoms was studied[1].

Application

Pentafluoropropionic acid (Perfluoropropionic acid) was used in the analysis of gentamicin with Gemini column by liquid chromatographic (LC) methods[2].
Pentafluoropropionic acid can be used to prepare 2-pentafluoropropylquinazolin-4(3H)-ones[3], perfluoroalkyl thioethers[4] and perfluoroalkyl-containing 3-azabicyclo[3.1.0]hexanes.[5]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective C-H Functionalization-Addition Sequence Delivers Densely Substituted 3-Azabicyclo [3.1. 0] hexanes
Pedroni, Julia and Cramer, Nicolai
Journal of the American Chemical Society, 139(36), 12398-12401 (2017)
One-Pot Synthesis of Trifluoromethylated Quinazolin-4 (3 H)-ones with Trifluoroacetic Acid as CF3 Source
Almeida, Sofia and Marti, Roger and Vanoli, Ennio and Abele, Stefan and Tortoioli, Simone
The Journal of Organic Chemistry, 83(9), 5104-5113 (2018)
E S Vasiliev et al.
The journal of physical chemistry. A, 118(23), 4013-4018 (2014-05-14)
The kinetics of the reaction between fluorine atoms and pentafluoropropionic acid has been studied experimentally at T = 262-343 K. The overall reaction rate constant decreases with temperature: k1(T) = 6.1 × 10(-13) exp(+1166 K)/T) cm(3) molecule(-1) s(-1). The potential
Synthesis of perfluoroalkyl thioethers from aromatic thiocyanates by iron-catalysed decarboxylative perfluoroalkylation
Exner, Benjamin and Bayarmagnai, Bilguun and Matheis, Christian and Goossen, Lukas J
Journal of Fluorine Chemistry, 198, 89-93 (2017)
B Li et al.
Journal of pharmaceutical and biomedical analysis, 55(1), 78-84 (2011-02-15)
Liquid chromatography combined with pulsed electrochemical detection (LC-PED) is the method of choice in the European Pharmacopoeia for the determination of gentamicin and its related substances. A recently approved improved LC-PED method, with a reversed-phase C(18) column and a mobile

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service