Skip to Content
Merck
All Photos(1)

Key Documents

345700

Sigma-Aldrich

Ganciclovir

A nucleoside analog structurally related to Acyclovir.

Synonym(s):

Ganciclovir, 9-[(1,3-Dihydroxy-2-propoxy)methyl]guanine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$306.00

$306.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$306.00

About This Item

Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$306.00


Estimated to ship on26 March 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

0.1 M HCl: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]1(c2[nH]c(n[c](c2nc1)=O)N)COC(CO)CO

InChI

1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)

InChI key

IRSCQMHQWWYFCW-UHFFFAOYSA-N

General description

A nucleoside analog structurally related to Acyclovir (Cat. No. 114798). Acts as a prodrug that is activated by phosphorylation. Has been used in the study of “suicide” gene therapy in cancer research. Upon expression of a viral suicide gene encoding herpes simplex virus, thymidine kinase (TK), the non-toxic prodrug ganciclovir, is converted to an active phosphorylated analog that can be incorporated into the DNA of replicating eukaryotic cells, causing death of the malignant dividing cell. Causes an irreversible cell cycle arrest at the G2/M checkpoint. Has also been used to study the loss of telomeres and to evaluate sensitivity of viruses to antiviral agents.
A nucleoside analog that is structurally related to Acyclovir (Cat. No. 114798). Acts as a pro-drug that is activated by phosphorylation. It has been used in the study of "suicide gene therapy" in cancer research. Upon expression of a viral suicide gene encoding herpes simplex virus thymidine kinase (TK), the non-toxic pro-drug is converted to the phosphorylated active analog and is incorporated into the DNA of replicating eukaryotic cells, causing death of the dividing malignant cells. Causes an irreversible cell cycle arrest at the G2/M checkpoint. Has also been used to study the loss of telomers and to evaluate the sensitivity of viruses to anti-viral agents.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Acts as a prodrug that is activated by phosphorylation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Aghi, M., et al. 2000. J. Gene Med.2, 148.
Qiao, J., et al. 2000. Hum. Gene Ther.11, 1569.
Cannon, J.S., et al. 1999. J. Virol.73, 4786.
Rubsam, L.Z., et al. 1999. Cancer Res.59, 669.
Sprung, C.N., et al. 1999. Proc. Natl. Acad. Sci. USA96, 6781.
Yamasaki, H., et al. 1999. C.R. Acad. Sci. III322, 151.
Halloran, P.J., and Fenton, R.G. 1998. Cancer Res.58, 3855.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco M Barriga et al.
Nature protocols, 19(5), 1381-1399 (2024-02-08)
The elimination of large genomic regions has been enabled by the advent of site-specific nucleases. However, as the intended deletions get larger, the efficiency of successful engineering decreases to a point where it is not feasible to retrieve edited cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service