Skip to Content
Merck
All Photos(2)

Key Documents

857135C

Avanti

18:1 BMP (S,S)

sn-(3-oleoyl-2-hydroxy)-glycerol-1-phospho-sn-1′-(3′-oleoyl-2′-hydroxy)-glycerol (ammonium salt), chloroform

Synonym(s):

lysobisphosphatidic acid; LBPA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$666.00

$666.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$666.00

About This Item

Empirical Formula (Hill Notation):
C42H82NO10P
CAS Number:
Molecular Weight:
792.07
UNSPSC Code:
51191904
NACRES:
NA.25

$666.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (BMP (contains 30% positional isomers), TLC)

form

liquid

packaging

pkg of 1 × 1 mL (857135C-10mg)
pkg of 1 × 1 mL (857135C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857135C

concentration

10 mg/mL (857135C-10mg)
5 mg/mL (857135C-5mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](COP(OC[C@@]([H])(O)COC(CCCCCCC/C=C\CCCCCCCC)=O)([O-])=O)(O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C42H79O10P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-35-39(43)37-51-53(47,48)52-38-40(44)36-50-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);1H3/b19-17-,20-18-;/t39-,40-;/m0./s1

InChI key

YRFISMFFADMBIY-BXURRMQSSA-N

Application

18:1 bis(monoacylglycerol)phosphate (BMP) (S,S) may be used as an standard in liquid chromatography-mass spectrometry (LC/MS) analysis for the quantification of BMP in mice tissues.[1]

Packaging

5 mL Clear Glass Sealed Ampule (857135C-10mg)
5 mL Clear Glass Sealed Ampule (857135C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic disease and ABHD6 alter the circulating bis (monoacylglycerol) phosphate profile in mice and humans
Grabner GF, et al.
Journal of Lipid Research, 60(5), 1020-1031 (2019)
Elena Zaitseva et al.
PLoS pathogens, 6(10), e1001131-e1001131 (2010-10-16)
Many enveloped viruses invade cells via endocytosis and use different environmental factors as triggers for virus-endosome fusion that delivers viral genome into cytosol. Intriguingly, dengue virus (DEN), the most prevalent mosquito-borne virus that infects up to 100 million people each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service