Skip to Content
Merck
All Photos(1)

Key Documents

344300

Sigma-Aldrich

sFRP-1 Inhibitor

The sFRP-1 Inhibitor, also referenced under CAS 915754-88-0, controls the biological activity of sFRP-1. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

sFRP-1 Inhibitor, Wnt Pathway Activator IV, N-(3-(Dimethylamino)propyl)-2-ethyl-5-(phenylsulfonyl)benzenesulfonamide, Secreted Frizzled Related Protein-1 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$219.00

$219.00


Estimated to ship on19 May 2025



Select a Size

Change View
5 MG
$219.00

About This Item

Empirical Formula (Hill Notation):
C19H26N2O4S2
CAS Number:
Molecular Weight:
410.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$219.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥95% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NCCCN(C)C)c1c(ccc(c1)[S](=O)(=O)c2ccccc2)CC

InChI

1S/C19H26N2O4S2/c1-4-16-11-12-18(26(22,23)17-9-6-5-7-10-17)15-19(16)27(24,25)20-13-8-14-21(2)3/h5-7,9-12,15,20H,4,8,13-14H2,1-3H3

InChI key

BHFFSIFXFHJJNJ-UHFFFAOYSA-N

General description

A diphenylsulfone-sulfonamide compound that counteracts the antagonizing effect of SARP-2/sFRP-1 (secreted frizzled related protein-1) against Wnt-3/frizzled interaction and Wnt-3/frizzled-stimulated cellular functions (EC50 = 1.27 µM in a U2OS-based reporter assay) with a reported aqueous solubility of 26 µg/ml at pH 7.4 and in vitro half-lives of 12 and 26.7 min using rat and human microsomes, respectively.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gopalsamy, A., et al. 2008. J. Med. Chem.51, 7670.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service