Skip to Content
Merck
All Photos(1)

Key Documents

21545

Millipore

IHC Select Citrate Buffer pH 6.0, 10x

Synonym(s):

Antigen/ Epitope Retrieval Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$186.00

$186.00


Estimated to ship on16 April 2025



Select a Size

Change View
250 ML
$186.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405

$186.00


Estimated to ship on16 April 2025


form

liquid

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

shipped in

wet ice

General description

For use as a Pretreatment Reagent in Immunohistochemical staining procedures after dilution. Use prepared 1X Citrate Buffer to retrieve Epitopes in formalin-fixed, paraffin-embedded tissue sections mounted on glass microscope slides. This procedure is performed after deparaffinization and prior to IHC1,2.

Application

Prediluted for use in Immunohistochemical staining procedures. For example:

Blocking Reagent

Primary Antibody or Negative Control Reagent

Biotinylated Secondary Antibody

Streptavidin-Enzyme Conjugate

Chromogen

Counterstain

For best results use this reagent in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.
Research Category
All

Physical form

Liquid Citrate Buffer containing 0.1% ProClin 300 as a preservative.

Storage and Stability

When stored at 2-8°C, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service