Skip to Content
Merck
All Photos(1)

Key Documents

16-204

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, recombinant clone 4G10®, biotin conjugate

clone 4G10®, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$908.00

$908.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$908.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$908.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PID1(55022)

General description

Included Positive Antigen Control: Catalog # 12-302, EGF-stimulated A431 cell lysate

Product Description: Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Heavy chain C-terminus has a hexa-histidine tag for purification and immobilization via nickel affinity matrices. Cross-linked to biotin. Patent pending.
Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Heavy chain C-terminus has a hexa-histidine tag for purification and immobilization via nickel affinity matrices. Cross-linked to biotin. Patent pending.

Specificity

Tyrosine-phosphorylated proteins from all species.

Immunogen

Phosphotyramine-KLH

Application

Detect Phosphotyrosine using this Anti-Phosphotyrosine Antibody, recombinant clone 4G10, biotin conjugate validated for use in WB.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Routinely evaluated by western blot to detect tyrosine phosphorylated proteins in a RIPA lysate of EGF-stimulated A431 carcinoma cells.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected tyrosine phosphorylated proteins in a RIPA lysate of EGF-stimulated A431 carcinoma cells.

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

Biotin-conjugated recombinant mouse IgG2bκ in PBS containing 0.05% sodium azide. Liquid at 2-8°C.
Protein G Purified

Storage and Stability

Stable for 9 months at 2-8ºC from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap. DO NOT FREEZE!

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin C Yarrow et al.
Chemistry & biology, 12(3), 385-395 (2005-03-31)
Small-molecule kinase inhibitors are predominantly discovered in pure protein assays. We have discovered an inhibitor of Rho-kinase (ROCK) through an image-based, high-throughput screen of cell monolayer wound healing. Using automated microscopy, we screened a library of approximately 16,000 compounds finding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service