Skip to Content
Merck
All Photos(1)

Key Documents

1.59007

Supelco

Acetic Acid Solution

0.1% (v/v) Acetic acid in water, hypergrade for LC-MS LiChrosolv®

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$117.00

$117.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
$117.00

About This Item

UNSPSC Code:
41116026
NACRES:
NA.03
technique(s):
HPLC: suitable
LC/MS: suitable
bp:
100 °C/1013 hPa

$117.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Water with 0.1% (v/v) Acetic acid, hypergrade for LC-MS LiChrosolv®

Quality Level

product line

LiChrosolv®

form

liquid

concentration

0.095-0.105 % (v/v)

technique(s)

HPLC: suitable
LC/MS: suitable

bp

100 °C/1013 hPa

storage temp.

2-30°C

General description

hypergrade for LC-MS LiChrosolv®

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service