Skip to Content
Merck
All Photos(1)

Key Documents

51197

Supelco

Sodium formate solution

suitable for LC-MS, LiChropur

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$343.00

$343.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$343.00

About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3723428
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21

$343.00


Check Cart for Availability

Request a Bulk Order

quality

LiChropur

concentration

10 mM NaOH in isopropanol/water 1:1 (+0.2% HCOOH)

technique(s)

LC/MS: suitable

suitability

suitable for LC-MS

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium formate solution is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS).
Features:       
  • Extensive QC testing ensures the highest specifications.
  • Optimized to improve ionization and resolution.    
  • Extremely low levels of inorganic and organic impurities.   
  • Manufactured specifically for accurate and fast LC-MS.

Application

Sodium formate may be used as a mobile phase additive in the following:        
  • enantiomeric separation of acidic compounds by nano-liquid chromatography (Nano-LC) technique[1]
  • determination of ambrisentan enantiomers in commercial samples of active pharmaceutical ingredients (API) using high-performance liquid chromatography (HPLC) technique[2]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of ambrisentan enantiomers by enantioselective liquid chromatography using cellulose?based chiral stationary phase in reverse phase mode
Dousa M and Gibala P
Journal of Separation Science, 35(7), 798-803 (2012)
Anna Rocco et al.
Journal of separation science, 32(10), 1696-1703 (2009-04-17)
Some racemic nonsteroidal anti-inflammatory drugs, namely naproxen, indoprofen, ketoprofen, flurbiprofen, carprofen, cicloprofen, flunoxaprofen and suprofen were separated into their enantiomers by nano-LC. Chiral recognition was achieved adding to the mobile phase heptakis (2,3,6-tri-O-methyl)-beta-cyclodextrin (TM-beta-CD). Capillary columns of 100 microm id

Questions

  1. What is the concentration of this sodium formate solution (51197 - 100 ml ) ?

    1 answer
    1. This product contains 0.2 wt. % formic acid and 0.04 wt. % of 10 mM Sodium Hydroxide in a 1:1 mixture of isopropanol and water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service