Skip to Content
Merck
All Photos(1)

Key Documents

1.52270

Supelco

Chromolith® WP 300 RP-18 (monolithic) HPLC Columns

L × I.D. 100 mm x 4.6 mm HPLC Column

Synonym(s):

Chromolith® WP 300 RP-18 100-4.6 HPLC column

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$1,860.00

$1,860.00


Estimated to ship on26 May 2025



Select a Size

Change View
1 UNIT
$1,860.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,860.00


Estimated to ship on26 May 2025


Product Name

Chromolith® WP 300 RP-18, L × I.D. 100 mm x 4.6 mm HPLC column

material

PEEK column

Quality Level

Agency

suitable for USP L1

product line

Chromolith®

feature

endcapped: no
endcapped: no

packaging

1 ea of

extent of labeling

18% Carbon loading
18% loading

parameter

200 bar pressure
200 bar pressure
60 °C max. temp.
60 °C max. temp.

technique(s)

HPLC: suitable
HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)
mass spectrometry (MS): suitable

column L × I.D.

10 cm × 4.6 mm

surface area

300 m2/g

matrix

monolithic silica
monolithic silica

matrix active group

C18 bonding phase
C18 bonding phase

pore size

2 μm macroporosity
2 μm macroporosity
30 mm mesoporosity
30 nm mesoporosity

operating pH range

1.5-7.5
2-7.5

separation technique

reversed phase

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

Biotherapeutics, for example, bio-engineered drugs and peptide therapeutics, represent the promise of new medical treatments for the future. Production costs have been falling leading to extremely high demand for suitable analytical methods for process monitoring and quality control of these biomolecules. HPLC is the preferred method of analysis, and it is therefore important to use the right column for these larger molecules.

Accurate analysis of proteins, antibodies, and large peptides requires columns with good permeability, along with better mass transfer and selectivity. In order for size-exclusion not to influence the separation, the pore size should be approximately ten times larger than the molecule being analyzed.

Chromolith columns have shown great potential and superiority in comparison to standard silica. Chromolith® WP 300 columns are made of monolithic silica with a characteristic bimodal pore structure.

Chromolith® WP 300 RP-18 columns have macropores with 2 μm diameter, giving a column efficiency exceeding 80,000 plates/meter. The mesopores are 30 nm (300 Å), and the surface modification is Octadecylsilane.

Chromolith® WP 300 RP-18 columns fall under the USP L1 Classification. The column efficiency is comparable to a 5 μm particle size fully porous particulate column. Chromolith® WP 300 RP-18 columns are suitable for LC-MS applications.The column dimension of 100 mm in length and 4.6 mm I.D. is the best choice for fast separations of Biomolecules on any HPLC System.Chromolith® WP 300 RP-18 100-4.6 HPLC columns enable:
  • Completely bioinert column hardware and high biorecovery
  • High-speed separations at very low column back-pressure
  • High matrix tolerance, allowing time and cost savings with simplified workflows
  • Cost savings due to extended column lifetimes
  • Connection of columns, flow gradients, and the use of mobile phases with higher viscosity
The recommended Guard-columns are: Chromolith® WP 300 RP-18, 5-4.6 guard cartridges (3 pieces) [1.52273] with the Chromolith® Guard Cartridge Holder Bioinert 5 mm x 4.6 mm [1.52255]Or Chromolith® WP 300 RP-18 10-4.6 guard cartridges (3 pieces) [1.52272] with the Chromolith® Guard Cartridge Holder Bioinert 10 mm x 4.6 mm [1.52256]. The Chromolith® column coupler can be used to significantly increase the separation efficiency of Chromolith® columns by coupling several columns in series. [1.51467] Learn more: Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns. Chromolith® WP 300 RP-18 columns are also available with 2 mm inner dimension. Flyer: Rapid and Robust Protein Chromatography - Chromolith® WP 300 RP-18 2 mm I.D. HPLC Columns. All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments. Supelco HPLC Products: The Best Choice for Any LC Instrument

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

Analytical Chromolith® HPLC columns for Bio applications, 300A mesopores: RP-18, RP-8, RP-4, Prot-A, Epoxy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service