Skip to Content
Merck
All Photos(1)

Key Documents

1.51467

Supelco

Chromolith® HPLC Column Coupler

Column coupler

Synonym(s):

Chromolith® column coupler for 10-32 female ports

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$120.00

$120.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 UNIT
$120.00

About This Item

UNSPSC Code:
41115716
NACRES:
SB.52

$120.00


Estimated to ship on01 June 2025


Product Name

Chromolith® Column Coupler, column coupler

Quality Level

product line

Chromolith®

feature

endcapped: no

technique(s)

HPLC: suitable

matrix

highly porous

storage temp.

no temp limit

General description

The Chromolith® HPLC column coupler is designed for linking several monolithic columns together in order to further increase separation efficiency and column performance. The combination results in a theoretical plate count that is significantly higher than any particulate column available. At the same time, pressure is kept well below the HPLC system limit.

Learn more:
Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns

All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument

Analysis Note

visual control: passes test

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Monolithic silica HPLC columns enable high-throughput analysis at low back pressure without loss of separation efficiency or peak capacity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service