Skip to Content
Merck
All Photos(1)

Key Documents

1.46704

Millipore

Sabouraud Dextrose Agar (selective) Settle Plate

with Lecithin, Tween®, histidine and sodium thiosulfate - ICR+ irradiated, 90 mm triple packed

Synonym(s):

SDA, ICR plus settle plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
Pricing and availability is not currently available. Contact Technical Service

Quality Level

material

polystyrene plate

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code
ready-to-use

D

90 mm

application(s)

air monitoring
microbiology
pharmaceutical

compatibility

for use with Air and personnel monitoring

storage temp.

15-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar + LTHTh - ICR in 90 mm settle plate is a selective media for the determination of the total count of yeasts and molds in air via active or passive air monitoring as well for fingerprints of personnel. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favors fungal growth and an antibacterial agent in the media inhibits the growth of bacterial flora. Neutralizers such as lecithin, polysorbate (Tween®) 80, histidine and sodium thiosulfate inactivate residues of disinfectants. In addition, ICRplus plates (plus or “+”) are supplied with a lockable lid for a safe transport after sampling without the risk of losing the lid. ICR plus plates are suitable for anaerobic or microaerophilic incubation of the plates, by turning the lid into the vent position

Application

Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service